Skip to Content
Merck
All Photos(1)

Documents

GE27-9557-01

illustra PuReTaq RTG PCR, 0.2 ML MWP

Cytiva 27955701, solid

Synonym(s):

PCR beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
55101520
NACRES:
NA.55

form

solid

manufacturer/tradename

Cytiva 27955701

storage condition

(Ambient temperature in pouch with desiccant)

color

white

General description

PuReTaq Ready-To-Go PCR Beads are pre-mixed, predispensed, single-dose reactions optimized for performing standard PCR amplifications. The use of recombinant PuReTaq DNA polymerase and other high-purity reagents ensures reliable and robust performance in both end point and real-time fluorescence-based PCR amplifications, and ensures the lowest possible levels of contaminating prokaryotic and eukaryotic nucleic acids.

The puReTaq Ready-To-Go PCR Beads are pre-formulated to ensure greater reproducibility between reactions, minimize pipetting steps, and reduce the potential for pipetting errors and contamination. The only additional reagents required are water, primers, and template DNA. The beads are provided pre-dispensed into either 0.2 ml or 0.5 ml PCR tubes. The 0.2 ml tubes are also supplied in a 96-well (8 × 12) plate format that allows individual strips of eight tubes to be easily removed. This flexibility allows use of either the entire 96-well plate, strips of eight, or individual 0.2 ml tubes.

Features and Benefits

Pre-formulated and pre-dispensed, freeze-dried PCR reagents mix in a bead format for robust and reproducible performance in standard PCR amplifications.

Save time: simply add template DNA solution and primers, and cycle.
  • More reproducible results: less risk of pipetting errors and contamination.
  • High-quality puReTaq DNA polymerase and high-purity reagents: robust performance and the lowest possible levels of contaminating DNA.
  • Long-term ambient-temperature stable: no freezer space required; less energy consumption for shipping and storage.
  • Verified for real-time PCR.
  • For highly robust and reproducible performance in standard PCR applications.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service