Skip to Content
Merck
All Photos(1)

Key Documents

GE27-0843-01

PreScission Protease

Cytiva 27-0843-01

Synonym(s):

fusion protein, GST-tag removal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106501
NACRES:
NA.56

packaging

pkg of 500 units

manufacturer/tradename

Cytiva 27-0843-01

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

PreScission Protease is a fusion protein of human rhinovirus (HRV) 3C protease and GST. It allows for on-column cleavage of GST tags and protein purification in one step.

On-column or off-column enzyme cleavage

GST tags and protease can be removed in a single step while fusion proteins are still bound to Glutathione Sepharose 4B, Glutathione Sepharose High Performance, or Glutathione Sepharose 4 Fast Flow affinity resin. Another option is to elute the GST-tagged protein using reduced glutathione, cleave, then pass the sample over the same resin to remove the protease and tags. Unlike other common proteases, such as thrombin and Factor Xa, this GST-tagged fusion protein can be removed easily through binding to a resin with affinity for GST. Other non-recombinant proteases require an additional purification step with a different resin.

Vectors encoding the HRV 3C protease recognition sequence

pGEX-6P vectors pGEX-6P-1, pGEX-6P-2, and pGEX-6P-3 encode the recognition sequence for this 3C protease.

Application

Specific, low temperature cleavage of fusion protein tags; Simple on-column cleavage during affinity purification
PreScission Protease has been used for eluting GST-tagged AT3, during in vitro ubiquitination reactions and AT3-Ub preparation. It has also been used to cleave GST tag from the a-enolase during induction of GST a-enolase expression in the protease-deficient BL21 strain of Escherichia coli.

Features and Benefits

  • Specific cleavage – between the Gln and Gly residues of the recognition sequence LeuGluValLeuPheGln/GlyPro.
  • Time-saving – GST-tagged proteins can be cleaved while still bound to an affinity resin.
  • Low incubation temperature – enables low temperature cleavage of fusion proteins containing the HRV 3C protease recognition sequence.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

pGEX Vectors
pGEX Vectors are to be used for scientific investigation and research and for no other purpose whatsoever and a license for commercial use of the licensed products and the processes claimed in US patent 5,654,176 and equivalent patents and patent applications in other countries must be negotiated directly with Millipore Corp (formerly Chemicon International Inc) by the purchaser prior to such use.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Wang et al.
Nature communications, 10(1), 3162-3162 (2019-07-20)
HECT E3 ligases control the degradation and functioning of numerous oncogenic/tumor-suppressive factors and signaling proteins, and their activities must be tightly regulated to prevent cancers and other diseases. Here we show that the Nedd4 family HECT E3 WWP1 adopts an
Merel A T van de Plassche et al.
Chembiochem : a European journal of chemical biology, 21(23), 3383-3388 (2020-07-28)
The global pandemic caused by SARS-CoV-2 calls for the fast development of antiviral drugs against this particular coronavirus. Chemical tools to facilitate inhibitor discovery as well as detection of target engagement by hit or lead compounds from high-throughput screens are
Thi-Bich Luu et al.
The New phytologist, 235(5), 1995-2007 (2022-05-26)
Rhizobial lipochitooligosaccharidic Nod factors (NFs), specified by nod genes, are the primary determinants of host specificity in the legume-Rhizobia symbiosis. We examined the nodulation ability of Medicago truncatula cv Jemalong A17 and M. truncatula ssp. tricycla R108 with the Sinorhizobium
Ji-Young Lee et al.
Cell reports, 27(12), 3602-3617 (2019-06-20)
The hepatitis C virus (HCV) is a major cause of chronic liver disease, affecting around 71 million people worldwide. Viral RNA replication occurs in a membranous compartment composed of double-membrane vesicles (DMVs), whereas virus particles are thought to form by
Dorothée Raoux-Barbot et al.
PloS one, 13(11), e0206133-e0206133 (2018-11-13)
Several bacterial pathogens produce nucleotidyl cyclase toxins to manipulate eukaryotic host cells. Inside host cells they are activated by endogenous cofactors to produce high levels of cyclic nucleotides (cNMPs). The ExoY toxin from Pseudomonas aeruginosa (PaExoY) and the ExoY-like module

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service