Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5080-01

HiTrap® rprotein A Fast Flow

Cytiva 17-5080-01, pack of 1 × 5 mL

Synonym(s):

HiTrap rProtein, Protein A Fast Flow Column, rProtein A Fast Flow

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

recombinant protein A (E. coli)

packaging

pack of 1 × 5 mL

manufacturer/tradename

Cytiva 17-5080-01

storage condition

(20% Ehtanol)
(20% Ethanol)

parameter

42 psi

bed size

16 mm × 25 mm

bed volume

5 mL

column I.D.

16 mm

matrix

4% cross-linked agarose

particle size

60-165 μm

average diameter

90 μm (d50v)

cleaning

2-11

working range

3-10

capacity

~50 mg binding capacity (human lgG/ml medium)

suitability

suitable for bioprocess medium

storage temp.

2-8°C

General description

Capacity of HiTrap rProtein A for some monoclonal antibodies. Running conditions: Binding buffer: 20 mM sodium phosphate (incl. 3 M NaCl for IgG), pH 7.0. Elution buffer: 0.1 M sodium citrate, pH 3.0.
HiTrap® rProtein A FF columns are prepacked with rProtein A Sepharose Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources, including cell supernatants and ascites.

The recombinant protein A ligand (produced in E. coli) is coupled to Sepharose Fast Flow by a technique which generates a stable thioether linkage between rProtein A and the base matrix. The coupling technique is optimized to give high binding capacity for IgG.
pH below 3 is sometimes required to elute strongly bound IgG species. However, protein ligands may hydrolyze at very low pH.

Features and Benefits

  • Quick, convenient preparative purification of monoclonal antibodies.
  • Well-established protein A-based medium used in many approved MAb processes.
  • Recombinant protein A exhibits similar Fc region specificity to that of native protein A but shows enhanced binding capacity and fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification.
  • Simple operations with a syringe, pump, or high-performance chromatography system, such as AKTA design.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service