Skip to Content
Merck
All Photos(1)

Documents

GE17371601

Capto MMC ImpRes, 25 ML

Cytiva 17371601, 40 μm avg. part. size (median particle size of the cumulative volume distribution)

Synonym(s):

Capto MMC ImpRes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

ligand

(Multimodal weak anion exchanger)

manufacturer/tradename

Cytiva 17371601

matrix

high-flow agarose

avg. part. size

40 μm (median particle size of the cumulative volume distribution)

cleaning in place

2-14

working range

3-12

capacity

60-90 mg Mab/ml binding capacity

Looking for similar products? Visit Product Comparison Guide

General description

The weak cation exchange multimodal ligand enables high selectivity in a broad pH/salt window compared with traditional ion exchangers, which allows the use of Capto MMC ImpRes in a variety of process conditions to solve challenging purification problems.

Features and Benefits

Capto MMC ImpRes is a weak cation exchange multimodal BioProcess chromatography medium (resin), designed to meet the demands of modern large-scale manufacturers, for high-resolution polishing of monoclonal antibodies (MAbs) and other biomolecules.

  • High yields achieved through the high-resolution beads and selectivity of the ligand.
  • Efficient removal of aggregates, viruses, and main contaminants in MAb processes.
  • Enables use of a platform approach to MAb process development.
  • Displays a broad pH/salt operational window.
  • Suitable for polishing of antibody fragments.

Storage and Stability

Store at 4 to 30 °C (20% ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service