GE17-0061-01
Sephadex® G-100 Superfine
Cytiva 17-0061-01, pack of 100 g
Synonym(s):
Size Exclusion Resin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
23151817
NACRES:
NA.56
Recommended Products
feature
autoclavable In wet form (pH 7.0) at 120°C for 30 min
packaging
pack of 100 g
manufacturer/tradename
Cytiva 17-0061-01
parameter
flow rate
matrix
cross-linked dextran
particle size
10-40 μm
25-100 μm (wet)
working range
2-10
Related Categories
General description
Sephadex® G-100 Superfine is useful for desalting and buffer exchange of very large biomolecules, for specific cases requiring very high resolution.
Application
Sephadex® is a gel filtration medium prepared by crosslinking Dextran with epichlorohydrin. Different types of Sephadex® differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex® G-10 is one of five different G-types ranging from G-10 for small molecules to G-75 for larger molecules. Sephadex® G-50 is available in 4 different particle sizes (Course, Medium, Fine & Superfine) and Superfine has the smallest bead size for higher efficiency with shorter diffusion distances. Cource and Medium are are preferred for large scale group separations where high flow rates and Low operating pressures are required.
Features and Benefits
- Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
- Classic gel filtration medium.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
Sephadex is a registered trademark of Cytiva
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service