GE10600000
Amersham™ Protran® Western blotting membranes, nitrocellulose
pore size 0.1 μm, roll W × L 300 mm × 4 m, pkg of 1 ea
Synonym(s):
Nitrocellulose membrane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 10600000
roll W × L
300 mm × 4 m
pore size
0.1 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
A versatile nitrocellulose blotting membrane with 0.1 µm pore size. Amersham Protran offers excellent sensitivity, resolution and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western blotting systems.
Application
Amersham™ Protran® Western blotting membranes, nitrocellulose is suitable for use in:
- Western blotting
- Southern blotting
- Northern blotting
Features and Benefits
- Low background and excellent signal-to-noise ratios.
- Compatible with a variety of labeling and detection systems.
- High binding capacity of small proteins (< Mr 10 000) reduces risk of "blow-through".
- Proprietary nitrocellulose formula gives a long shelf life of bound proteins.
- Available in popular precut sizes for laboratory application.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service