Skip to Content
Merck
All Photos(2)

Key Documents

GBL247880

Grace Bio-Labs ProPlate® tray sets

size 16 wells, round, with Delrin® snap clips

Synonym(s):

multi array slide system, multi slide module system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122600
NACRES:
NB.15
material:
black
round wells
silicone (on Delrin)
size:
16 wells

material

black
round wells
silicone (on Delrin)

sterility

non-sterile

manufacturer/tradename

Grace Bio-Labs 247880

size

16 wells

well area

0.28 cm2

well volume

350 μL

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Grace Bio-Labs ProPlate® multi-array chamber tray sets are designed to be used with ONCYTE® nitrocellulose-coated slides. These multi-array chambers integrate microscope slide based microarray technology with high-throughput (HTS applications) microtiter plate processing. Up to 64 individual arrays can be assayed per slide.

Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.

Features:

  • Up to 16 or 64 leak-proof wells on any slide surface without the use of adhesive
  • Microtiter plate well spacing facilitates use with multichannel pipettes
  • 7 x 7 mm square wells are ideal for high content arrays 5-350 uL well volumes accommodate small reagent and large wash volumes

16 or 64 well ProPlate® tray sets include the following:

  • 7 x 7 mm well dimensions (x 4)
  • 16 or 64 well capacity
  • Stainless steel Delrin® snap clips (x 8)
  • Tray & cover included (x 1)
  • 10 seal strips (x 10) and 1 seal strip applicator (x 1)

Legal Information

Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service