Skip to Content
Merck
All Photos(1)

Key Documents

G9144

Sigma-Aldrich

Anti-Gliadin (Wheat) antibody produced in rabbit

fractionated antiserum, buffered aqueous solution

Synonym(s):

Anti-Gliadin (Wheat)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

wheat

technique(s)

dot blot: 1:1,500
indirect ELISA: 1:5,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Gliadin is a protein found in wheat and other cereals. The T cells recognize the gliadin epitopes and activate the innate immune response similar to the response to pathogens. This response is more intense in coeliac disease wherein the gut-derived T cells produced in response to gliadin induce damage to the small intestine. Therefore, patients affected by coeliac disease or gluten-sensitivity should avoid food that produce gliadin
Gliadin is an important component of wheat gluten. It is composed of single-chain polypeptides and has a molecular weight of 25−100 kDa. It is associated by intramolecular disulfide bonds.

Specificity

Rabbit Anti-Gliadin shows specificity for native wheat gliadin.

Immunogen

native and heat-treated wheat gliadin

Application

Anti-Gliadin (Wheat) antibody produced in rabbit has been used in immunoblot and enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Gliadin is a class of glutamine containing, alcohol soluble proteins separable from wheat and rye glutens known as the prolamins. These proteins are associated with the harmful effects of celiac disease and gluten sensitive enteropathy in humans, causing characteristic changes in the small intestinal mucosa. In such cases, strict compliance to a gluten-free diet is usually recommended.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Garcia-Calvo et al.
Foods (Basel, Switzerland), 10(1) (2021-01-06)
Gluten is the ethanol-soluble protein fraction of cereal endosperms like wheat, rye, and barley. It is widely used in the food industry because of the physical-chemical properties it gives to dough. Nevertheless, there are some gluten-related diseases that are presenting
Determination of Prolamins in Beers by ELISA and SDS-PAGE
Kanerva P, et al.
Journal of the Institute of Brewing, 111(1), 61-64 (2005)
Luigi Maiuri et al.
Lancet (London, England), 362(9377), 30-37 (2003-07-11)
The adaptive immune system is central to the development of coeliac disease. Adaptive immune responses are, however, controlled by a preceding activation of the innate immune system. We investigated whether gliadin, a protein present in wheat flour, could activate an
Tobias L Freitag et al.
Gastroenterology, 158(6), 1667-1681 (2020-02-08)
Celiac disease could be treated, and potentially cured, by restoring T-cell tolerance to gliadin. We investigated the safety and efficacy of negatively charged 500-nm poly(lactide-co-glycolide) nanoparticles encapsulating gliadin protein (TIMP-GLIA) in 3 mouse models of celiac disease. Uptake of these
The Level of Expression of Thioredoxin is Linked to Fundamental Properties and Applications of Wheat Seeds
Li YC, et al.
Molecular plant, 2(3), 430-441 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service