Skip to Content
Merck
All Photos(1)

Key Documents

G8162

Sigma-Aldrich

β-Glucuronidase from Escherichia coli

aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)

Synonym(s):

β-D-Glucuronide glucuronosohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Escherichia coli

Quality Level

form

aqueous glycerol solution

specific activity

≥5,000,000 units/g protein, pH 6.8 (biuret)

mol wt

69-71 kDa

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Glucuronidase from Escherichia coli is similar to human glucuronidase enzyme and corresponds to molecular weight close to 69-71 kDa and has an pH optimum of 6.5-7.5. It belongs to family-2 glycosyl hydrolase and has active site residues glutamic acid 394, tyrosine 468 and glutamic acid 504.

Application

β-Glucuronidase from Escherichia coli has been used in the enzymatic cleavage and activation of glucuronide prodrugs in non-small cell lung cancer cells, U87 human glioblastoma cell line, in A549 (human lung adenocarcinoma) and KB (human oral squamous carcinoma).
β-Glucuronidase from E. coli is used for the enzymatic hydrolysis of b-glucuronides in urine and other fluids. It has a high rate of hydrolytic activity and may be useful for determining the presence of androsterone, 17-hydroxycorticosteroids, and estriol in urine.
The optimal conditions for the enzymatic hydrolysis of α-hydroxytriazolam, one of the major metabolites of triazolam in human urine, were determined using β-glucuronidase Type IX-A.
It is used as a reporter gene in GUS assays to monitor gene expression.
New Technical Article Comparing Performance of Different Enzymes
Learn more
about recent application data generated by Sigma R&D to optimize hydrolysis for different drug classes using enzymes from different sources and the use of a chromatographicaly purified enzyme to reduce the effect of esterase activity resulting in conversion of 6-MAM to Morphine
Effective in the hydrolysis of steroid glucuronides.
Used for the hydrolysis of glucuronide conjugates in urinary metabolite analysis

Biochem/physiol Actions

β-Glucuronidase catalyzes the hydrolysis of β-glucuronic acid residues from the non-reducing termini of glycosaminoglycans (GAGs). β-Glucuronidase is a potential candidate enzyme for gene-mediated enzyme prodrug therapy for glucuronide prodrugs. Use of β-Glucuronidase-albumin complex based drug delivery could be an effective therapeutic methodology for treating solid tumors.
β-glucuronidase (β-GIc) is an exoglycosidase that catalyzes the breakdown of complex carbohydrates. In humans it converts conjugated bilirubin into the unconjugated form, making bilirubin suitable for reabsorption.

Unit Definition

One Sigma or modified Fishman unit will liberate 1.0 μg of phenolphthalein from phenolphthalein glucuronide per hr at 37 °C at the pH 6.8 (30 min assay).

Physical form

Highly purified solution in 50% glycerol

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Membrane-localized activation of glucuronide prodrugs by beta-glucuronidase enzymes
Chen KC, et al.
Cancer Gene Therapy, 14(2), 187-187 (2007)
Mickaël Thomas et al.
Bioorganic & medicinal chemistry, 16(17), 8109-8116 (2008-08-12)
Two glucuronide prodrugs of the histone deacetylase inhibitor CI-994 were synthesized. These compounds were found to be soluble in aqueous media and stable under physiological conditions. The carbamoyl derivatisation of CI-994 significantly decreased its toxicity towards NCI-H661 lung cancer cells.
Synthesis and biological evaluations of a monomethylauristatin E glucuronide prodrug for selective cancer chemotherapy
Legigan T, et al.
European Journal of Medicinal Chemistry, 67, 75-80 (2013)
Expression and Purification of Escherichia coli beta-Glucuronidase
Aich S, et al.
Protein Expression and Purification, 22(1), 75-81 (2001)
Study of a cyclopamine glucuronide prodrug for the selective chemotherapy of glioblastoma
Hamon F, et al.
European Journal of Medicinal Chemistry, 45(4), 1678-1682 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service