Skip to Content
Merck
All Photos(1)

Key Documents

G7777

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)–Gold antibody produced in goat

affinity isolated antibody, aqueous glycerol suspension, 10 nm (colloidal gold)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

gold conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

aqueous glycerol suspension

particle size

10 nm (colloidal gold)

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases . Mouse IgGs have four distinct isotypes, namely, IgG1, IgG2a, IgG2b, and IgG3. IgG1 regulates complement fixation in mice . Anti-Mouse IgG (whole molecule)-Gold antibody is specific for mouse IgG.

Immunogen

Purified mouse IgG

Application

Anti-Mouse IgG (whole molecule)-Gold antibody is suitable for use in immunoelectron microscopy .

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Conjugate is suspended in Tris buffered saline, pH 8.2, containing 1% (w/v) BSA, 30% (v/v) glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihisa Hirakawa et al.
Journal of cell science, 125(Pt 24), 6176-6184 (2012-10-06)
In plants, many nucleus-encoded proteins are targeted to both mitochondria and plastids, and this process is generally mediated by ambiguous N-terminal targeting sequences that are recognized by receptors on both organelles. In many algae, however, plastids were acquired by secondarily
S B Shah et al.
Chromosoma, 105(2), 111-121 (1996-08-01)
Immunoelectron microscopy with anti-nucleolin defined substructures within the multiple nucleoli of biosynthetically active stage II-III oocytes and within the nucleoli of relatively quiescent stage VI oocytes of Xenopus laevis. Dense fibrillar components (DFCs) of nucleoli from stage II-III oocytes consisted
Maria Gregori et al.
Nanomedicine : nanotechnology, biology, and medicine, 13(2), 723-732 (2016-11-05)
Aggregation of amyloid-β peptide (Aβ) is a key event in the pathogenesis of Alzheimer's disease (AD). We investigated the effects of nanoliposomes decorated with the retro-inverso peptide RI-OR2-TAT (Ac-rGffvlkGrrrrqrrkkrGy-NH2) on the aggregation and toxicity of Aβ. Remarkably low concentrations of
Ke Wang et al.
Signal transduction and targeted therapy, 5(1), 283-283 (2020-12-06)
In face of the everlasting battle toward COVID-19 and the rapid evolution of SARS-CoV-2, no specific and effective drugs for treating this disease have been reported until today. Angiotensin-converting enzyme 2 (ACE2), a receptor of SARS-CoV-2, mediates the virus infection
Chongluo Fu et al.
Diseases of aquatic organisms, 65(1), 17-22 (2005-07-27)
Recently, an enveloped, spherical RNA virus was identified as the causative agent of mass mortalities among adult scallop Chlamys farreri, which is cultured on the northern coast of China. Hybridomas were prepared from mice immunized with highly purified virions. Four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service