Skip to Content
Merck

G702000

DMT-rG(tac)-CPG

pore size 1000 Å, 20-30 μmol/g

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
12352200
Assay:
≥98.0% (Nucleoside Purity, reversed phase HPLC)
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (Nucleoside Purity, reversed phase HPLC)

pore size

1000 Å

capacity

≥98 % coupling capacity (Coupling Efficiency)

storage temp.

−20°C



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library