Skip to Content
Merck
All Photos(1)

Key Documents

G6771

Sigma-Aldrich

Growth Hormone Releasing Factor Fragment 1-29 amide human

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C149H246N44O42S
CAS Number:
Molecular Weight:
3357.88
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](C)NC(=O)[C@@H](N)Cc1ccc(O)cc1)C(=O)N[C@@H](Cc2ccccc2)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc3ccc(O)cc3)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)NCC(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCCNC(N)=N)C(N)=O

InChI

1S/C149H246N44O42S/c1-20-77(13)116(191-122(211)81(17)168-132(221)104(66-113(204)205)178-121(210)79(15)167-123(212)88(152)62-84-39-43-86(198)44-40-84)145(234)185-102(63-83-32-23-22-24-33-83)138(227)193-118(82(18)197)146(235)186-103(65-111(155)202)137(226)189-108(71-196)142(231)182-101(64-85-41-45-87(199)46-42-85)136(225)175-93(38-31-56-165-149(161)162)126(215)174-91(35-26-28-53-151)131(220)190-115(76(11)12)143(232)184-97(58-72(3)4)124(213)166-68-112(203)170-94(47-49-109(153)200)128(217)180-100(61-75(9)10)135(224)188-106(69-194)140(229)169-80(16)120(209)172-92(37-30-55-164-148(159)160)125(214)173-90(34-25-27-52-150)127(216)179-99(60-74(7)8)134(223)181-98(59-73(5)6)133(222)176-95(48-50-110(154)201)129(218)183-105(67-114(206)207)139(228)192-117(78(14)21-2)144(233)177-96(51-57-236-19)130(219)187-107(70-195)141(230)171-89(119(156)208)36-29-54-163-147(157)158/h22-24,32-33,39-46,72-82,88-108,115-118,194-199H,20-21,25-31,34-38,47-71,150-152H2,1-19H3,(H2,153,200)(H2,154,201)(H2,155,202)(H2,156,208)(H,166,213)(H,167,212)(H,168,221)(H,169,229)(H,170,203)(H,171,230)(H,172,209)(H,173,214)(H,174,215)(H,175,225)(H,176,222)(H,177,233)(H,178,210)(H,179,216)(H,180,217)(H,181,223)(H,182,231)(H,183,218)(H,184,232)(H,185,234)(H,186,235)(H,187,219)(H,188,224)(H,189,226)(H,190,220)(H,191,211)(H,192,228)(H,193,227)(H,204,205)(H,206,207)(H4,157,158,163)(H4,159,160,164)(H4,161,162,165)/t77-,78-,79-,80-,81-,82+,88-,89-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,115-,116-,117-,118-/m0/s1

InChI key

WGWPRVFKDLAUQJ-MITYVQBRSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2

General description

GHRH/GRH (growth hormone releasing hormone) is an endocrine peptide hormone composed of 44 amino acids† It exists in two forms in the hypothalamus, namely GRH(1-44)-NH2 and GRH(1-40)-OH. However, the full biological functionality of this hormone resides in the first 29 amino acids itself. GRH(1-29)-NH2 is a C-terminal shortened peptide produced from GHRH.

Biochem/physiol Actions

Amidated GRF fragment that is equipotent to GRF in releasing somatotropin from anterior pituitary.
GHRH/GRH (growth hormone releasing hormone) is released from the hypothalamus from where it reaches the anterior pituitary and binds to its receptor, thus, eventually leading to the release of GH (growth hormone) into the plasma.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burcu Ayhan-Sahin et al.
PloS one, 17(1), e0260144-e0260144 (2022-01-22)
Growth Hormone Releasing Hormone (GHRH), 44 amino acids containing hypothalamic hormone, retains the biological activity by its first 29 amino acids. GHRH (NH2 1-29) peptide antagonists inhibit the growth of prostate, breast, ovarian, renal, gastric, pancreatic cancer in vitro and
Ferenc G Rick et al.
Cell cycle (Georgetown, Tex.), 11(22), 4203-4210 (2012-10-26)
Treatment of colon cancer with an antagonist of growth hormone-releasing hormone (GHRH), JMR-132, results in a cell cycle arrest in S-phase of the tumor cells. Thus, we investigated the effect of JMR-132 in combination with S-phase-specific cytotoxic agents, 5-FU, irinotecan
Lucie Jetté et al.
Endocrinology, 146(7), 3052-3058 (2005-04-09)
In vivo bioconjugation to the free thiol on Cys34 of serum albumin by a strategically placed reactive group on a bioactive peptide is a useful tool to extend plasma half-life. Three maleimido derivates of human GH-releasing factor (hGRF)(1-29) were synthesized
L A Frohman et al.
The Journal of clinical investigation, 83(5), 1533-1540 (1989-05-01)
The plasma enzyme responsible for primary proteolytic cleavage of growth hormone-releasing hormone (GRH) at the 2-3 amino acid bond was characterized. Native GRH[GRH(1-44)-NH2 and GRH(1-40)-OH], and COOH-terminally shortened fragments [GRH(1-32)-NH2 and GRH(1-29)-NH2] were rapidly cleaved, while GRH(2-32)-NH2 was not degraded
Laura Muñoz-Moreno et al.
International journal of cancer, 132(4), 755-765 (2012-07-11)
New approaches are needed to the therapy of advanced prostate cancer. This study determined the effect of growth hormone-releasing hormone (GHRH) antagonists, JMR-132 and JV-1-38 on growth of PC3 tumors as well as on angiogenesis and metastasis through the evaluation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service