Skip to Content
Merck
All Photos(1)

Key Documents

G5922

Sigma-Aldrich

Anti-GW182 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-TNRC6A, Anti-Trinucleotide repeat containing 6A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 2.5-5.0 μg/mL using human epithelial HEp-2 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

GW182 is a phosphorylated cytoplasmic autoantigen. It contains multiple glycine-tryptophan (GW) repeats along with a single RNA recognition motif. GW182 is a protein of 182 kDa, found in mammalian cell types.

Application

Anti-GW182 antibody produced in rabbit has been used as a primary antibody for immunostaining of HeLa cells and human hepatoma cells. It has also been used for indirect immunofluorescence at a working concentration of 2.5-5.0μg/mL using human epithelial HEp-2 cells.

Biochem/physiol Actions

Glycine-tryptophan (GW) bodies or P-bodies are cytoplasmic bodies that are involved in mRNA degradation, storage and translational repression. These bodies contain Argonaute proteins and m-RNAs that associate with GW182. GW182 is involved in post-transcriptional gene silencing/RNA interference (RNAi) via the microRNA pathway. Inhibition of gene expression leads to destruction of GWBs and disruption of RNAi and microRNA-induced gene silencing as it forms an integral component of GWBs.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew Jakymiw et al.
Journal of cell science, 120(Pt 8), 1317-1323 (2007-04-03)
GW bodies, also known as mammalian P-bodies, are cytoplasmic foci involved in the post-transcriptional regulation of eukaryotic gene expression. Recently, GW bodies have been linked to RNA interference and demonstrated to be important for short-interfering-RNA- and microRNA-mediated mRNA decay and
A phosphorylated cytoplasmic autoantigen, GW182, associates with a unique population of human mRNAs within novel cytoplasmic speckles
Eystathioy T, et al.
Molecular Biology of the Cell, 13(4), 1338-1351 (2002)
GW182 is critical for the stability of GW bodies expressed during the cell cycle and cell proliferation
Yang Z, et al.
Journal of Cell Science, 117(23), 5567-5578 (2004)
Terence N Bukong et al.
Hepatology (Baltimore, Md.), 57(1), 70-80 (2012-08-18)
Alcohol use and hepatitis C virus (HCV) infection synergize to cause liver damage, and microRNA-122 (miR-122) appears to play a key role in this process. Argonaute 2 (Ago2), a key component of the RNA-induced silencing complex (RISC), has been shown
Jidong Liu et al.
Nature cell biology, 7(12), 1261-1266 (2005-11-15)
In animals, the majority of microRNAs regulate gene expression through the RNA interference (RNAi) machinery without inducing small-interfering RNA (siRNA)-directed mRNA cleavage. Thus, the mechanisms by which microRNAs repress their targets have remained elusive. Recently, Argonaute proteins, which are key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service