Skip to Content
Merck
All Photos(5)

Key Documents

G5009

Sigma-Aldrich

γ-Globulins from bovine blood

≥99% (agarose gel electrophoresis)

Synonym(s):

Bovine γ-Globulin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

bovine blood

Quality Level

Assay

≥99% (agarose gel electrophoresis)

form

powder

composition

NaCl, ≤4%

technique(s)

enzyme immunoassay: suitable

solubility

0.9% NaCl: soluble 25 mg/mL

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Bovine gamma globulin (BGG) is an important component of the immune system of cows and other bovines. BGG is found in the blood of bovines and is primarily used in the production of medical products such as vaccines, serums, and diagnostic reagents. BGG can also be used to treat certain medical conditions, such as hepatitis B, as well as to boost immunity in humans and other animals.

Application

Bovine γ-globulins were used to study the constituents and pH changes in protein rich hyaluronan solution which affect the biological properties of artificial articular joints. It is also used to determine immunoglobulin G in bovine colostrum and milk using direct biosensor SPR-immunoassay.
Bovine γ-globulin has been used as an immunological challenge in studies of mechanisms of immunotolerance.

Biochem/physiol Actions

The γ-G globulins of bovine serum are categorized by anion exchange chromatography, immunoelectrophoresis, zone electrophoresis, ultracentrifugation, and analysis of the products of papain digestion. They have properties similar to analogous components of human serum. This was determined by the methods such as: gel filtration, immunoelectrophoresis, anion exchange chromatography, ultracentrifugation, and reduction with mercaptoethanol. Fast and slow γ-G globulins were classified on the basis of differences in electrophoretic migration rates, chromatographic elution positions, and biological activities (complement fixation). Bovine γ-A was conditionally found in immunoelectrophoretic analyses of serum and chromatographic fractions.

Preparation Note

Prepared from Cohn Fraction II, III

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Fang et al.
eLife, 11 (2022-01-21)
The ring-like ATPase complexes in the AAA+ family perform diverse cellular functions that require coordination between the conformational transitions of their individual ATPase subunits (Erzberger and Berger, 2006; Puchades et al., 2020). How the energy from ATP hydrolysis is captured
Marc-Olivier Montjovent et al.
Bioanalysis, 9(18), 1385-1393 (2017-09-29)
Recombinant glycoprotein produced in nonhuman mammalian cell lines can be modified with the immunogenic nonhuman sialic N-glycolylneuraminic acid (Neu5Gc). We describe here a validated method for detection of antidrug antibodies against both protein and Neu5Gc-containing glycan epitopes. An electrochemiluminescent method
Harvey E Indyk et al.
Journal of AOAC International, 86(2), 386-393 (2003-05-02)
An automated biosensor surface-plasmon resonance-based assay was developed for the determination of immunoglobulin G (IgG) in bovine milk and colostrum with either goat or rabbit antibovine IgG or protein G used as detecting molecule. The method is configured as a
T Kitano et al.
Journal of biomechanics, 34(8), 1031-1037 (2001-07-13)
The relationship between the coefficient of friction and pH value or protein constituents of lubricating fluid, together with viscosity, were studied within a bearing surface model for artificial joint, ultra-high molecular weight polyethylene (UHMWPE) against stainless steel (SUS), using a
Zijing Chen et al.
Genetics, 215(3), 713-728 (2020-05-22)
TRP channels function in many types of sensory receptor cells. Despite extensive analyses, an open question is whether there exists a family of auxiliary subunits, which could influence localization, trafficking, and function of TRP channels. Here, using Drosophila melanogaster, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service