Skip to Content
Merck
All Photos(2)

Key Documents

G1508

Sigma-Aldrich

Glycogen from mussel, Mytilus genus

≥85% anhydrous basis (enzymatic)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

(Mytilus genus)

Assay

≥85% anhydrous basis (enzymatic)

form

powder

impurities

≤1% Protein (Lowry)

color

white to off-white

storage temp.

2-8°C

InChI

1S/C24H42O21/c25-1-5-9(28)11(30)16(35)22(41-5)39-4-8-20(45-23-17(36)12(31)10(29)6(2-26)42-23)14(33)18(37)24(43-8)44-19-7(3-27)40-21(38)15(34)13(19)32/h5-38H,1-4H2/t5-,6-,7-,8-,9-,10-,11+,12+,13-,14-,15-,16-,17-,18-,19-,20-,21+,22+,23-,24-/m1/s1

InChI key

BYSGBSNPRWKUQH-UJDJLXLFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycogen is a branched polymer of glucose synthesized by animal cells for energy storage and release. It is constructed of predominantly α1→4 glycosidic bonds with branches created through α1→6 glycosidic bonds.

Application

Glycogen, a starch analogue, is added to polyacrylamide gels for starch phosphorylase activity aβays were it acts as a substrate for starch phosphorylase degradation.

Preparation Note

Prepared by a modification of the procedure of Bell, et al., Biochem. J., 28, 882 (1934).

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne Borland et al.
Journal of experimental botany, 57(2), 319-328 (2005-12-17)
In the halophytic species Mesembryanthemum crystallinum, crassulacean acid metabolism (CAM) may be induced by a range of abiotic factors including drought, salinity, high light intensity, low temperature, and anoxia. A key biotic consequence of all these environmental changes is the
Katsiaryna Skryhan et al.
PloS one, 10(9), e0136997-e0136997 (2015-09-15)
Starch biosynthesis in Arabidopsis thaliana is strictly regulated. In leaf extracts, starch synthase 1 (AtSS1) responds to the redox potential within a physiologically relevant range. This study presents data testing two main hypotheses: 1) that specific thiol-disulfide exchange in AtSS1
Rebecca I Clark et al.
Cell, 155(2), 435-447 (2013-10-01)
Infections disturb metabolic homeostasis in many contexts, but the underlying connections are not completely understood. To address this, we use paired genetic and computational screens in Drosophila to identify transcriptional regulators of immunity and pathology and their associated target genes
Malou Friederich-Persson et al.
Hypertension (Dallas, Tex. : 1979), 62(5), 914-919 (2013-09-11)
Diabetic nephropathy is strongly associated with both increased oxidative stress and kidney tissue hypoxia. The increased oxidative stress causes increased kidney oxygen consumption resulting in kidney tissue hypoxia. To date, it has been difficult to determine the role of kidney
Monica Dus et al.
Nature neuroscience, 16(5), 526-528 (2013-04-02)
Animals can determine the nutritional value of sugar without the influence of taste. We examined a Drosophila mutant that is insensitive to the nutritional value of sugars, responding only to the concentration (that is, sweetness). The affected gene encodes a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service