Skip to Content
Merck
All Photos(1)

Documents

G0776

Sigma-Aldrich

Disialoganglioside-GD2 from bovine brain

~95%, lyophilized powder, semisynthetic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

~95%

form

lyophilized powder

storage temp.

−20°C

InChI

1S/C74H134N4O32/c1-4-6-8-10-12-14-16-18-19-21-23-25-27-29-31-33-52(89)78-43(44(84)32-30-28-26-24-22-20-17-15-13-11-9-7-5-2)40-101-69-61(95)60(94)63(50(38-81)104-69)106-70-62(96)67(64(51(39-82)105-70)107-68-55(77-42(3)83)59(93)58(92)49(37-80)103-68)110-74(72(99)100)35-46(86)54(76)66(109-74)57(91)48(88)41-102-73(71(97)98)34-45(85)53(75)65(108-73)56(90)47(87)36-79/h30,32,43-51,53-70,79-82,84-88,90-96H,4-29,31,33-41,75-76H2,1-3H3,(H,77,83)(H,78,89)(H,97,98)(H,99,100)/b32-30+/t43?,44?,45-,46-,47?,48?,49-,50-,51-,53+,54+,55-,56?,57?,58+,59-,60-,61-,62-,63-,64+,65+,66+,67-,68+,69-,70+,73-,74+/m1/s1

InChI key

FFILOTSTFMXQJC-QCFYAKGBSA-N

Amino Acid Sequence

Cer-Glc-Gal(NeuAc-NeuAc)-GalNAc

General description

Disialoganglioside GD2 is a cell surface-associated glycosphingolipid, which comprises sialic acid moiety and is regarded as a carbohydrate antigen.. It is expressed in the skin melanocytes, central and peripheral nervous system.
Gangliosides are major constituents of neuronal cell membranes and endoplasmic reticulum; contain a sialated polysaccharide chain linked to ceramide through a β-glycosidic linkage; for classification of gangliosides see Svennerholm, L., et al. (eds.), Structure and Function of Gangliosides, New York, Plenum, 1980.

Application

Disialoganglioside-GD2 from bovine brain has been used:
  • in the ganglioside- enzyme-linked immunosorbent assay (ELISA)
  • in capture ELISA with anti-GD2 antibody (ch14.18)
  • as a control to test its effect on reactivating autophagy in primary fibroblasts

Biochem/physiol Actions

Disialoganglioside GD2 is expressed in neuroblastoma, melanoma and small cell lung cancer. It is an immunotherapeutic target and is also regarded as a tumor-associated antigen. GD2 may favor cell migrationin tumors by promoting cell growth, as well as extracellular matrix component attachment.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A E Druĭ et al.
Voprosy onkologii, 58(4), 514-520 (2012-01-01)
The bone marrow (BM) TH, ELAVL4 and GD2 genes expression was evaluated in 331 samples from 57 different stage neuroblastoma (NB) patients, 26 BM samples from patients without NB and samples from 2 NB cell lines (IMR-32, Kelly) by real-time
Daisuke Sakai et al.
Nature communications, 3, 1264-1264 (2012-12-13)
Despite the high prevalence of intervertebral disc disease, little is known about changes in intervertebral disc cells and their regenerative potential with ageing and intervertebral disc degeneration. Here we identify populations of progenitor cells that are Tie2 positive (Tie2+) and
Wenyong Tong et al.
Methods in molecular biology (Clifton, N.J.), 928, 39-52 (2012-09-08)
Ganglioside GD2 is a cell surface glycosphingolipid that is targeted clinically for cancer diagnosis, prognosis, and therapy. The conformations of free GD2 and of GD2 bound to anti-GD2 mAb 3F8 were resolved by saturation transfer difference nuclear magnetic resonance and
Marie Vincent et al.
International journal of cancer, 133(3), 757-765 (2013-01-29)
Immunocytokines (ICKs) targeting cytokines to the tumor environment using antibodies directed against a tumor-associated antigen often have a higher therapeutic index than the corresponding unconjugated cytokines. Various ICKs displaying significant antitumoral effects in several murine tumor models have already been
S L Pichla et al.
Journal of structural biology, 119(1), 6-16 (1997-06-01)
The GD2 ganglioside is a cell-surface component that appears on the surface of metastatic melanoma cells and is a marker for the progression of the disease. The ME36.1 monoclonal antibody binds to the GD2 ganglioside and has shown potential as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service