Skip to Content
Merck
All Photos(2)

Key Documents

G0513

Sigma-Aldrich

Gly-Pro p-nitroanilide hydrochloride

≥99% (HPLC)

Synonym(s):

GP-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N4O4 · HCl
CAS Number:
Molecular Weight:
328.75
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥99% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, faintly yellow to yellow

shipped in

wet ice

storage temp.

−20°C

SMILES string

Cl.NCC(=O)N1CCCC1C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C13H16N4O4.ClH/c14-8-12(18)16-7-1-2-11(16)13(19)15-9-3-5-10(6-4-9)17(20)21;/h3-6,11H,1-2,7-8,14H2,(H,15,19);1H

InChI key

VBBFIBMVFSUUBP-UHFFFAOYSA-N

Application

Gly-Pro p-nitroanilide hydrochloride has been used as a substrate in dipeptidyl peptidase-IV (DPP-IV) enzyme assay. It has also been used to determine DPP-IV inhibitory activity.

Substrates

Substrate for Dipeptidyl Peptidase IV.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monitoring of dipeptidyl peptidase-IV (DPP-IV) activity in patients with mucopolysaccharidoses types I and II on enzyme replacement therapy?Results of a pilot study
Hetmanczyk K, et al.
Clinical Biochemistry, 49(6), 458-462 (2016)
Multifunctional biological activities of water extract of housefly larvae (Musca domestica)
Li H, et al
PharmaNutrition, 5(4), 119-126 (2017)
Chemical composition and inhibitory activities on dipeptidyl peptidase IV and pancreatic lipase of two underutilized species from the Brazilian Savannah: Oxalis cordata A. St.-Hil. and Xylopia aromatica (Lam.) Mart
Oliveira VB, et al.
Food Research International, 105, 989-995 (2018)
Antidiabetic evaluation of Hemionitis arifolia leaves by in vitro methods
Kumar KKR, et al.
Manipal journal of pharmaceutical sciences., 1(1), 13-20 (2015)
Emily Mason et al.
Foods (Basel, Switzerland), 9(7) (2020-07-18)
The bioactive properties and health-promoting effects of two novel yellow (C09052, C05041) and two brown (Calvi, Bastia) hairless canary seed (Phalaris canariensis L.) cultivars were investigated in comparison to two common cereal grains (wheat and oat). The cereal flours were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service