Skip to Content

F9803

FTI-277 trifluoroacetate salt

≥95% (HPLC), film

Synonym(s):

N-[4-[2(R)-Amino-3-mercaptopropyl]amino-2-phenylbenzoyl]methionine methyl ester trifluoroacetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H29N3O3S2 · xC2HF3O2
CAS Number:
Molecular Weight:
447.61 (free base basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

film

storage condition

desiccated

solubility

H2O: ≥2 mg/mL

shipped in

dry ice

storage temp.

−70°C

SMILES string

OC(=O)C(F)(F)F.COC(=O)[C@H](CCSC)NC(=O)c1ccc(NC[C@@H](N)CS)cc1-c2ccccc2

InChI

1S/C22H29N3O3S2.C2HF3O2/c1-28-22(27)20(10-11-30-2)25-21(26)18-9-8-17(24-13-16(23)14-29)12-19(18)15-6-4-3-5-7-15;3-2(4,5)1(6)7/h3-9,12,16,20,24,29H,10-11,13-14,23H2,1-2H3,(H,25,26);(H,6,7)/t16-,20+;/m1./s1

InChI key

GJEFFRDWFVSCOJ-PXPMWPIZSA-N

Application

Farnesyltransferase inhibitor 277 (FTI-277) has been used to:
  • inhibit protein farnesylation in breast cell lines.
  • in marrow-isolated adult multilineage inducible cells (MIAMI).
  • inhibition of farnesyl transferase in CV-1 in Origin with SV40 genes cells (COS-7).

Biochem/physiol Actions

Highly potent (pM/nM) Ras CAAX peptidomimetic which antagonizes both H and K-Ras oncogenic signaling. Inhibitor of farnesyltransferase (Ftase) IC50 = 50 nM.
Farnesyltransferase inhibitor 277 (FTI-277) mediates apoptosis in multiple myeloma and is regarded as a potential therapeutic agent.

Packaging

Very hygroscopic material, package in a dry room with deoxygenated MeOH and pump down the solvent. Store with desiccant.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library