Skip to Content
Merck
All Photos(3)

Key Documents

F9677

Sigma-Aldrich

Felodipine

solid

Synonym(s):

4-(2,3-Dichlorophenyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinecarboxylic acid ethyl methyl ester, Plendil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19NO4Cl2
CAS Number:
Molecular Weight:
384.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

solubility

DMSO: 20 mg/mL
H2O: insoluble

originator

AstraZeneca

SMILES string

CCOC(=O)C1=C(C)NC(C)=C(C1c2cccc(Cl)c2Cl)C(=O)OC

InChI

1S/C18H19Cl2NO4/c1-5-25-18(23)14-10(3)21-9(2)13(17(22)24-4)15(14)11-7-6-8-12(19)16(11)20/h6-8,15,21H,5H2,1-4H3

InChI key

RZTAMFZIAATZDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Felodipine is a dihydropyridine derivative. It has diuretic properties. Felodipine has high vascular selectivity. It is used to treat hypertension. Felodipine regulates chronic stable angina.

Biochem/physiol Actions

L-type calcium channel blocker

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2 Inhalation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruixin Ma et al.
Journal of hypertension, 30(1), 210-216 (2011-12-03)
To evaluate the impact of felodipine with irbesartan on sexual function compared with felodipine with metoprolol in hypertensive women. This was a prospective, randomized, parallel, active-controlled, open-label study (ClinicalTrials.org: NCT01238705) in 160 women (18-60 years) with mild or moderate hypertension
Haiyan Li et al.
International journal of pharmaceutics, 427(2), 270-275 (2012-03-01)
Tomographic imaging techniques are attractive tools for the visualization of the internal structural characteristics of pharmaceutical solid dosage forms. In this paper, the internal structure of the tablet core for a monolith osmotic drug delivery system, felodipine sustained-release tablet, was
Morten Schmidt et al.
European journal of clinical investigation, 42(3), 266-274 (2011-08-13)
The CYP3A4 inhibition by calcium channel blockers (CCBs) may attenuate the effectiveness of clopidogrel. Using time-varying drug exposure ascertainment, we examined whether CCB use modified the association between clopidogrel use and major adverse cardiovascular events (MACE) after coronary stent implantation.
Yuqing Zhang et al.
Journal of hypertension, 30(11), 2202-2212 (2012-09-20)
In antihypertensive treatment trials, when randomized therapies do not reach target, additional drugs are administered. However, patients requiring (add-on) or not requiring add-on therapy (no-add-on) may be at different cardiovascular risk and differently susceptible to benefits of antihypertensive treatment. The
Effects of felodipine on haemodynamics and exercise capacity in patients with angina pectoris.
Sheridan JV, et al.
British Journal of Clinical Pharmacology, 23(4), 391-396 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service