Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C18H18N2S·HBr
CAS Number:
Molecular Weight:
375.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Quality Segment
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
light yellow to yellow
solubility
DMSO: ≥10 mg/mL
storage temp.
2-8°C
SMILES string
[s]1c(nc(c1)c3ccc(cc3)C)c2cc(ncc2)CCC.Br
InChI
1S/C18H18N2S.BrH/c1-3-4-16-11-15(9-10-19-16)18-20-17(12-21-18)14-7-5-13(2)6-8-14;/h5-12H,3-4H2,1-2H3;1H
InChI key
RJCFNQZVFUMORB-UHFFFAOYSA-N
General description
Fatostatin is a non-sterol diarylthiazole derivative. It prevents insulin-induced adipogenesis and lowers the amounts of fatty acid, triglyceride and low-density lipoprotein. Fatostatin has anti tumor and antimitotic properties.
Application
Fatostatin hydrobromide has been used:
- to study its anti-cancer activity and effects on mitotic microtubule spindle
- to study its effects on stomatal development
- to prevent SREBP cleavage-activating protein (SCAP)-mediated escort of sterol regulatory element-binding proteins (SREBPs)
Biochem/physiol Actions
Fatostatin hydrobromide is an SREBP inhibitor.
Fatostatin hydrobromide is an SREBP inhibitor. Fatostatin hydrobromide inhibits fat production and lowers glucose levels in mice by inhibiting SREBP (Sterol Regulatory Element Binding Proteins).
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.