Skip to Content
Merck
All Photos(3)

Key Documents

F8514

Sigma-Aldrich

Flurbiprofen

cyclooxygenase inhibitor

Synonym(s):

(±)-2-Fluoro-α-methyl-4-biphenylacetic acid, L-790,330

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H3(F)CH(CH3)CO2H
CAS Number:
Molecular Weight:
244.26
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76

biological source

synthetic

Quality Level

Assay

≥98.5% (HPLC)

form

powder

color

white to off-white

mp

110-112 °C (lit.)

solubility

methanol: 50 mg/mL

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

SMILES string

CC(C(O)=O)c1ccc(c(F)c1)-c2ccccc2

InChI

1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)

InChI key

SYTBZMRGLBWNTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Flurbiprofen is a nonsteroidal anti-inflammatory agent (NSAIA) with antipyretic, analgesic, and antifungal activity. Oral formulations of flurbiprofen may be used to treat rheumatoid arthritis, osteoarthritis and anklylosing spondylitis. Flurbiprofen is also used topically during ocular surgery to prevent or reduce intraoperative miosis. Flurbiprofen is structurally and pharmacologically related to ibuprofen. It is used to study the pathophysiological steps of NSAID enteropathy in the rat and the biotransformation of flurbiprofen by Cunninghamella species.

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor; non-steroidal anti-inflammatory agent with antifungal activity. The S enantiomer inhibits prostaglandin synthesis and has both anti-inflammatory and analgesic activity, while the R enantiomer does not inhibit prostaglandin synthesis and displays only analgesic activity.
Fluibiprofen is a cyclooxygenase (COX) inhibitor, which is an enzyme responsible for the conversion of arachidonic acid to prostaglandin G2 (PGG2) and PGG2 to prostaglandin H2 (PGH2) in the prostaglandin synthesis pathway. This decreases the prostaglandins which cause inflammation, pain, swelling and fever. Flurbiprofen inhibits the activity of both COX-1 and -2. The S enantiomer inhibits prostaglandin synthesis and has both anti-inflammatory and analgesic activity, while the R enantiomer does not inhibit prostaglandin synthesis and displays only analgesic activity.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica Amadio et al.
Applied and environmental microbiology, 76(18), 6299-6303 (2010-07-27)
The biotransformation of the fluorinated anti-inflammatory drug flurbiprofen was investigated in Cunninghamella spp. Mono- and dihydroxylated metabolites were detected using gas chromatography-mass spectrometry and fluorine-19 nuclear magnetic resonance spectroscopy, and the major metabolite 4'-hydroxyflurbiprofen was isolated by preparative high-pressure liquid
T Mahmud et al.
Gut, 43(6), 775-782 (1998-11-21)
Non-steroidal anti-inflammatory drugs (NSAIDs) cause gastrointestinal damage by a non-prostaglandin (PG) dependent "topical" action and by inhibiting cyclooxygenase. To discriminate between these two effects by studying some key pathophysiological steps in NSAID enteropathy following administration of (R)- and (S)-flurbiprofen, the
N M Davies
Clinical pharmacokinetics, 28(2), 100-114 (1995-02-01)
Flurbiprofen is a chiral nonsteroidal anti-inflammatory drug (NSAID) of the 2-arylpropionic acid class. Although it possesses a chiral centre, with the S-(+)-enantiomer possessing most of the beneficial anti-inflammatory activity, both enantiomers may possess analgesic activity and all flurbiprofen preparations to
Mirco Govoni et al.
The Journal of pharmacology and experimental therapeutics, 346(1), 96-104 (2013-04-19)
Liver first-pass metabolism differs considerably among organic nitrates, but little information exists on the mechanism of denitration of these compounds in hepatic tissue. The metabolism of nitrooxybutyl-esters of flurbiprofen and ferulic-acid, a class of organic nitrates with potential therapeutic implication
Structural and functional basis of cyclooxygenase inhibition.
Anna L Blobaum et al.
Journal of medicinal chemistry, 50(7), 1425-1441 (2007-03-08)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service