Skip to Content
Merck
All Photos(1)

Key Documents

F8190

Sigma-Aldrich

Fucoidan from Fucus vesiculosus

≥95%

Synonym(s):

Fucoidin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

algae (Fucus Vesiculosus)

Assay

≥95%

form

powder

color

light yellow

useful pH range

5.0-11.0

solubility

water: 10 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Fucoidan is a fucose-rich, sulfated polysaccharide isolated from several species of brown seaweed.

Application

Fucoidan from Fucus vesiculosus has been to:
  • study the application of heparin red in detecting fucoidan
  • study its bioactivity as an antimicrobial agent
  • study its effects on advanced glycation end-products

Biochem/physiol Actions

Studies of fucoidan have found it to have a broad variety of biological activities including antitumor and antiangogenic activities, immunomodulatory, anti-inflammatory, anticoagulant, antithrombotic, and antioxidant activities. Fucoidan induces macrophage activation with subsequently activates mitogen activated protein kinases (MAPKs) and results in natural killer cell proliferation. Fucoidan acts as a nonselective selectin blocker and has been shown to induce apoptosis and suppress angiogenesis.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of scavenger receptors-1 class A stimulation on macrophage morphology and highly modified advanced glycation end product-protein phagocytosis
Shinichi H et al.
Scientific Reports, 8(1), 5901-5901 (2018)
Fanlu Wang et al.
Marine drugs, 19(4) (2021-04-04)
Fucoidans, sulfated polysaccharides extracted from brown algae, are marine products with the potential to modulate bone formation and vascularization processes. The bioactivity and safety of fucoidans are highly associated with their chemical structure, which may vary with algae species and
Bioactivity of Fucoidan as an Antimicrobial Agent in a New Functional Beverage
Poveda-Castillo et al.
Becker-Shaffer's Diagnosis and Therapy of the Glaucomas (2018)
Hang T T Cao et al.
Marine drugs, 16(11) (2018-11-06)
Fucoidans from brown macroalgae have beneficial biomedical properties but their use as pharma products requires homogenous oligomeric products. In this study, the action of five recombinant microbial fucoidan degrading enzymes were evaluated on fucoidans from brown macroalgae: Sargassum mcclurei, Fucus
Quantification of sulfated polysaccharides in mouse and rat plasma by the Heparin Red mix-and-read fluorescence assay
Ulrich W et al.
ARKIVOC (Gainesville, FL, United States) (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service