Skip to Content
Merck
All Photos(1)

Key Documents

F7887

Sigma-Aldrich

Anti-Bovine IgG (whole molecule)–FITC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Bovine IgG (whole molecule)–Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

direct immunofluorescence: 1:300
immunohistochemistry: 1:300

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Immunoglobulin G (IgG) contributes to 10-20% of plasma protein and is regarded as one of the most predominant serum protein. It consists of four subclasses: IgG1, IgG2, IgG3 and IgG4. The IgG structure possesses four polypeptide chains containing two identical γ heavy (H) chains and two identical κ or λ light (L) chains of 50 kDa and 25 kDa, respectively.

Application

Anti-Bovine IgG (whole molecule)-FITC antibody produced in rabbit has been used in indirect fluorescent antibody test (IFAT).

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of FITC to Anti- Bovine IgG (whole molecule) antibody allows for the visualization of protein by fluorescent microscopy.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Serological survey of Neospora caninum and Toxoplasma gondii in cattle (Bos indicus) and water buffaloes (Bubalus bubalis) in ten provinces of Brazil
Spagnol FH, et al.
Revista Brasileira de Parasitologia veterinaria = Brazilian Journal of Veterinary Parasitology : Org?o Oficial do Colegio Brasileiro de Parasitologia Veterinaria, 52, 42-45 (2009)
L D Monje et al.
Journal of medical entomology, 53(3), 660-665 (2016-01-23)
Several cases of human rickettsiosis caused by Rickettsia parkeri were recently documented in the Paraná River delta of Argentina, where the tick vector is Amblyomma triste Koch. As cattle suffer recurrent A. triste infestations, they are at risk of becoming
Patricio M Calandra et al.
Revista Argentina de microbiologia, 46(4), 315-319 (2015-01-13)
The aim of this study was to describe two events in which Neospora caninum was involved in bovine abortions in beef cattle. In the first event, 11 abortions in 57 heifers were recorded in 45 days. One aborted heifer was
Sero-epidemiological survey on toxoplasmosis in cattle, sheep and goats in Algeria
Dechicha AS, et al.
African Journal of Agricultural Research, 18(2), 2113-2119 (2015)
Karin Persson Waller et al.
Animals : an open access journal from MDPI, 13(2) (2023-01-22)
An outbreak of disease in a Swedish beef cattle herd initiated an in-depth study to investigate the presence of bacteria and viruses in the blood of clinically healthy (n = 10) and clinically diseased cattle (n = 20) using whole-genome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service