Skip to Content
Merck
All Photos(1)

Key Documents

F6261

Sigma-Aldrich

Anti-Guinea Pig IgG (whole molecule)−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Guinea Pig IgG (whole molecule)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct immunofluorescence: 1:64

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Goat polyclonal anti-Guinea Pig IgG (whole molecule)−FITC antibody reacts with guinea pig IgG (whole molecule). Identity and purity of the antibody is established by immunoelectrophoresis (IEP), prior to conjugation. Electrophoresis of the antibody preparation followed by diffusion versus anti-goat IgG and anti-goat whole serum results in single arcs of precipitation.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.
Anti-Guinea Pig IgG (whole molecule)-FITC antibody is specific for guinea pig IgG subclasses. Goat anti-Mouse IgG is purified by affinity isolation and conjugated to FITC.
Second antibodies (secondary antibodies) are antibodies raised against other antibodies, typically from another species, called primary antibodies. Primary antibodies are generated with and bind to specific antigens to create antigen antibody complexes which may be detected in a variety of immunochemical and immunohistochemical configurations involving soluble or insoluble signaling molecules. The immunodetection system depends upon the binding of a signal competent secondary antibody to a primary antibody:antigen complex.
The suitability of any secondary antibody to a specific immunodetection application is dependent upon its signal development system configuration. Signal development systems are based upon the chemical conjugation of a signal mediator to the secondary antibody. Major secondary antibody signal systems are color (chromogenic), chemiluminescence or fluorescence based and involve the use of enzymes or high affinity binary systems such as biotin:avidin.
Fluorescein isothiocyanate (FITC) is a fluorescein derivative (fluorochrome) used to tag antibodies, including secondary antibodies, for use in fluorescence-based assays and procedures. FITC excites at 495 nm and emits at 521 nm.

Immunogen

Purified guinea pig IgG

Application

Anti-guinea pig IgG (whole molecule)-FITC antibody may be used for immunofluorescence of guinea pig spleen cells at a minimum dilution of 1:64. Antibody concentrations ranging from 1:80-1:100 were used for immunofluorescence in various studies. Guinea pig IgG was used as secondary antibody to quantitate fluorescence by photon counting using a Leitz MPV Compact 2 microscope photometer.
Goat polyclonal anti-Guinea Pig IgG (whole molecule)−FITC antibody may be used as a secondary antibody or to detect the presence of guinea pig IgG via fluorogenic immunochemical or immunohistochemical techniques.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen Brosi et al.
Journal of immunology (Baltimore, Md. : 1950), 183(11), 7187-7195 (2009-11-06)
RIP-B7.1 mice express the costimulator molecule B7.1 (CD80) on pancreatic beta cells and are a well-established model for studying de novo induction of diabetogenic CD8 T cells. Immunization of RIP-B7.1 mice with preproinsulin (ppins)-encoding plasmid DNA efficiently induces experimental autoimmune
Gabriela M Soares et al.
Journal of cellular physiology, 233(9), 7112-7119 (2018-03-27)
GTPase activating proteins (GAPs) are ubiquitously expressed, and their role in cellular adhesion and membrane traffic processes have been well described. TBC1D1, which is a Rab-GAP, is necessary for adequate glucose uptake by muscle cells, whereas increased TCGAP, which is
P Hedlund et al.
British journal of pharmacology, 116(4), 2258-2266 (1995-10-01)
1. The distribution and effects of pituitary adenylate cyclase-activating polypeptide (PACAP-27 and -38), helospectin (Hel-1 and Hel-2), and vasoactive intestinal polypeptide (VIP), were investigated in isolated preparations of human corpus cavernosum (CC). 2. Immunohistochemistry revealed coinciding profiles of nerve structures
Stage-specific surface antigens of the cattle lungworm Dictyocaulus viviparus
Britton C et al
Pediatrics International : Official Journal of the Japan Pediatric Society, 15, 625-634 (1993)
Mattia Camera et al.
Frontiers in neuroscience, 16, 744693-744693 (2022-03-04)
p140Cap, encoded by the gene SRCIN1 (SRC kinase signaling inhibitor 1), is an adaptor/scaffold protein highly expressed in the mouse brain, participating in several pre- and post-synaptic mechanisms. p140Cap knock-out (KO) female mice show severe hypofertility, delayed puberty onset, altered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service