Skip to Content
Merck
All Photos(4)

Key Documents

F5130

Sigma-Aldrich

5-Fluorouridine

proapoptotic anitproliferative plant growth regulator

Synonym(s):

5-Fluorouracil 1β-D-ribofuranoside, FUrd

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11FN2O6
CAS Number:
Molecular Weight:
262.19
Beilstein:
33662
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

Assay

≥99% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N2C=C(F)C(=O)NC2=O

InChI

1S/C9H11FN2O6/c10-3-1-12(9(17)11-7(3)16)8-6(15)5(14)4(2-13)18-8/h1,4-6,8,13-15H,2H2,(H,11,16,17)/t4-,5-,6-,8-/m1/s1

InChI key

FHIDNBAQOFJWCA-UAKXSSHOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Fluorouridine (FUrd) is a fluoropyrimidine nucleoside analog and a cell-permeable modified RNA precursor.

Application

5-Fluorouridine has been used for labeling active transcription sites in the porcine fetal fibroblasts and human cell lines for immunocytochemistry analysis. It has also been used to monitor apoptosis during drug sensitivity assay in esophageal squamous cell carcinoma (ESCC) cells.

Biochem/physiol Actions

5-Fluorouridine (FUrd) is cytotoxic towards cancer cells. FUrd is often used in chemical and biochemical comparison studies with fluorouracil and thymine analogs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P V Sahasrabudhe et al.
Nucleic acids research, 23(19), 3916-3921 (1995-10-11)
The effects of 5-fluorouridine (FUrd) and 5-fluorodeoxyuridine (FdUrd) substitution on the stabilities of duplex RNA and DNA have been studied to determine how FUrd substitution in nucleic acids may alter the efficiency of biochemical processes that require complementary base pairing
Nikolaos Tsesmetzis et al.
Cancers, 10(7) (2018-07-26)
Antimetabolites, in particular nucleobase and nucleoside analogues, are cytotoxic drugs that, starting from the small field of paediatric oncology, in combination with other chemotherapeutics, have revolutionised clinical oncology and transformed cancer into a curable disease. However, even though combination chemotherapy
Inhibition of RNA synthesis by 5-fluorouridine accounts for its cyto toxicity on colorectal cancer cells in vitro
Subbarayan PR, et al.
Cancer research, 65(9) (2005)
Angelica M Bello et al.
Journal of medicinal chemistry, 52(6), 1648-1658 (2009-03-06)
A series of 6-substituted and 5-fluoro-6-substituted uridine derivatives were synthesized and evaluated for their potential as anticancer agents. The designed molecules were synthesized from either fully protected uridine or the corresponding 5-fluorouridine derivatives. The mononucleotide derivatives were used for enzyme
P M Ellery et al.
Placenta, 30(4), 329-334 (2009-02-14)
The aim was to test for evidence of transcriptional activity within the nuclei of the syncytiotrophoblast of the human placenta. The syncytiotrophoblast forms the epithelial covering of the villous tree, and is a multinucleated, terminally-differentiated syncytium generated through fusion of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service