Skip to Content
Merck
All Photos(1)

Key Documents

F3133

Sigma-Aldrich

Fibroblast Growth Factor from bovine pituitary

lyophilized, suitable for cell culture

Synonym(s):

FGF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

product name

Fibroblast Growth Factor from bovine pituitary, suitable for cell culture

biological source

bovine pituitary glands

Quality Level

form

lyophilized powder

potency

1-100 ng/mL

mol wt

16.4 kDa

packaging

pkg of 10 and 5X10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white to off-white

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Fibroblast growth factors (FGF) are glycoproteins concealed in the extracellular matrix and cell surface. There are around 22 different types of FGFs known. These factors weigh in the range of 17 to 34kDa and they possess a conserved sequence of 120 amino acids.

Application

A natural mitogenic growth factor used in cell culture applications to study cell signaling and to promote fibroblast and endothelial cell growth and survival.
Fibroblast Growth Factor from bovine pituitary has been used in in vitro testicular cell culture. It has also been used as a component of myogenic culture medium.

Biochem/physiol Actions

Fibroblast growth factors (FGF) actively participate in cellular responses including embryogenesis and organogenesis. It specifically controls cell proliferation, differentiation and migration. FGF is known to regulate nervous system functions, tissue repair mechanism and wound healing. It also controls angiogenesis of tumor cells. Abnormal FGF mediated signaling induces tumor angiogenesis, by stimulating cancer cell proliferation and survival. FGFs carries out its function by mediating through four receptor tyrosine kinases : FGF-receptors FGFR1, FGFR2, FGFR3 and FGFR4.

Quality

Although the major mitogenic component of this product is FGF2, the activity per unit weight of protein is less than that of purified FGF2.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 0.1 mg/ml BSA.

Preparation Note

Purified glands by a modification of the method of Gospodarowicz.

Analysis Note

The biological activity is measured using a 3-day MTT assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modulation of osteogenic, adipogenic and myogenic differentiation of mesenchymal stem cells by submicron grooved topography
Wang PY, et al.
Journal of Materials Science. Materials in Medicine, 23(12), 3015-3028 (2012)
Fibroblast growth factor signalling: from development to cancer
Turner N and Grose R
Nature Reviews. Cancer, 10(2), 116-116 (2010)
Identification, culture, and characterization of germline stem cell-like cells in chicken testes
Jung JG, et al.
Biology of Reproduction, 76(1), 173-182 (2007)
A testis-mediated germline chimera production based on transfer of chicken testicular cells directly into heterologous testes
Lee YM, et al.
Biology of Reproduction, 75(3), 380-386 (2006)
Characterization and In Vitro Differentiation of Korean Ring-Necked Pheasant (Phasianus colchicus) Male Germ Cells
Jeong DK, et al.
Journal of Embryology transfer , 29(4), 351-359 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service