Skip to Content
Merck
All Photos(1)

Key Documents

F2756

Sigma-Aldrich

Fluorescein di(β-D-galactopyranoside)

Synonym(s):

FDG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32O15
CAS Number:
Molecular Weight:
656.59
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NC.07
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

powder

solubility

methanol: 20 mg/mL, clear, yellow

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc3c(Oc4cc(O[C@@H]5O[C@H](CO)[C@H](O)[C@H](O)[C@H]5O)ccc4C36OC(=O)c7ccccc67)c2)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C32H32O15/c33-11-21-23(35)25(37)27(39)30(45-21)42-13-5-7-17-19(9-13)44-20-10-14(43-31-28(40)26(38)24(36)22(12-34)46-31)6-8-18(20)32(17)16-4-2-1-3-15(16)29(41)47-32/h1-10,21-28,30-31,33-40H,11-12H2/t21-,22-,23+,24+,25+,26+,27-,28-,30-,31-/m1/s1

InChI key

ZTOBILYWTYHOJB-WBCGDKOGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescein di(β-D-galactopyranoside) (FDG) has been used for the detection of gene expression via FACS (fluorescence-activated cell sorting).[1][2]
Fluorescein di(β-D-galactopyranoside) has been used as a substrate in β-galactosidase assay.[3]

Biochem/physiol Actions

Fluorescein di (β-D-galactopyranoside) (FDG) results in fluorescence after being hydrolyzed to fluorescein by β-galactosidase. It gives fluorescence proportional to the enzymatic activity. This molecule is mainly required for the detection of gene expression in transfected cells when using β-galactosidase (Lac Z) as a reporter gene.[4][5]

Substrates

Fluorogenic substrate for β-galactosidase

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jose M Anton-Rodriguez et al.
Otology & neurotology : official publication of the American Otological Society, American Neurotology Society [and] European Academy of Otology and Neurotology, 40(6), 826-835 (2019-04-30)
To investigate whether [F]fluorothymidine (FLT) and/or [F]fluorodeoxyglucose (FDG) positron emission tomography (PET) can differentiate growth in neurofibromatosis 2 (NF2) related vestibular schwannomas (VS) and to evaluate the importance of PET scanner spatial resolution on measured tumor uptake. Six NF2 patients
Dominik Berzaczy et al.
Cancers, 12(1) (2019-12-22)
The response evaluation criteria in lymphoma (RECIL) classification for lymphoma treatment response assessment was introduced in 2017, but it has not yet been compared to the established Lugano classification. Also, the value of the provisional "minor response" (MiR) category of
N K Grzimek et al.
Journal of virology, 73(6), 5043-5055 (1999-05-11)
Transcription of the major immediate-early (MIE) genes of cytomegaloviruses (CMV) is driven by a strong promoter-enhancer (MIEPE) complex. Transactivator proteins encoded by these MIE genes are essential for productive infection. Accordingly, the MIEPE is a crucial control point, and its
A fluorimetric method using fluorescein di-?-D-galactopyranoside for quantifying the senescence-associated ?-galactosidase activity in human foreskin fibroblast Hs68 cells.
Yang NC and Hu ML
Analytical Biochemistry, 325, 337-343 (2004)
Use of fluorescein-di-beta-D-galactopyranoside (FDG) and C12-FDG as substrates for beta-galactosidase detection by flow cytometry in animal, bacterial, and yeast cells.
Plovins A
Applied and Environmental Microbiology, 60, 4638-4638 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service