Skip to Content
Merck

Skip To

EPS003

CUDC 101

A potent inhibitor of HDACs and receptor tyrosine kinases

Synonym(s):

7-[[4-(3-Ethynylphenylamino)-7-methoxyquinazolin-6-yl]oxy]-N-hydroxyheptanamide, Heptanamide, 7-[[4-[(3-ethynylphenyl)amino]-7-methoxy-6-quinazolinyl]oxy]-N-hydroxy

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H26N4O4
CAS Number:
Molecular Weight:
434.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.41
Assay:
≥98% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

color

white

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

N(O)C(=O)CCCCCCOc1cc2c(ncnc2Nc3cc(ccc3)C#C)cc1OC

InChI

1S/C24H26N4O4/c1-3-17-9-8-10-18(13-17)27-24-19-14-22(21(31-2)15-20(19)25-16-26-24)32-12-7-5-4-6-11-23(29)28-30/h1,8-10,13-16,30H,4-7,11-12H2,2H3,(H,28,29)(H,25,26,27)

InChI key

PLIVFNIUGLLCEK-UHFFFAOYSA-N

General description

A potent multitargeted inhibitor of histone deacetylase (HDAC) and the receptor kinases epidermal growth factor receptor (EGFR) and human epidermal growth factor receptor 2 (HER2), with IC50 values of 4.4, 2.4, and 15.7 nM, respectively.

Displays potent antiproliferative and proapoptotic activities against cultured and implanted tumor cells that are sensitive or resistant to several approved single-targeted drugs.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2221C7249SML2850
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

DMSO: soluble

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: >10 mg/mL, H2O: >10 mg/mL

solubility

DMSO: 2 mg/mL, clear

color

white

color

white to beige

color

-

color

white to beige


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters