Key Documents
EP5427757007
Rotor for Eppendorf® 5430/5430R Centrifuges
S-24-11-AT, aerosol-tight, to hold, 24 x 1.5/2.0 mL tubes, with lid and wind shield
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 24 x 1.5/2.0 mL tubes
aerosol-tight
swing bucket rotor
with lid and wind shield
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5427757007 (Global)
parameter
12,700 rpm
16,050 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5430/5430R
General description
Rotors to fot Eppendorf centrifuge 5430 and 5430 R.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)