Key Documents
EP0030128591
Eppendorf® Twin tec PCR plates, 96 wells
green plate, conical bottom colorless wells, semi-skirted, pkg of 25 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
conical bottom colorless wells
green plate
polycarbonate
size:
96 wells
sterility:
non-sterile
feature:
skirt (Semi skirted)
material
conical bottom colorless wells
green plate
polycarbonate
sterility
non-sterile
feature
skirt (Semi skirted)
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030128591
technique(s)
PCR: suitable
size
96 wells
working volume
250 μL
General description
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Features and Benefits
  • Eppendorf® Twin tec PCR plate combine a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Certified free of any detectable human DNA, DNase, RNase and PCR inhibitors.
  • Fits most thermal cyclers.
  • 250 μL max. well volume (when used with cap strips).
  • Improved well-to-well tolerance.
  • Ideal for quantitative real-time PCR as well – OptiTrack® matrix for faster sample identification and fewer pipetting errors.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)