EP0030127706
Eppendorf® Serological Pipets, Sterile Grade
green, capacity 2 mL, pack of 600 (4 bags × 150 pcs.)
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.04
Recommended Products
material
green
sterility
sterile
feature
DNA free
graduations
plugged
packaging
pack of 600 (4 bags × 150 pcs.)
manufacturer/tradename
Eppendorf® 0030127706 (Global)
capacity
2 mL
General description
- Eppendorf® Serological Pipets are manufactured from ultra-pure virgin polystyrene for optimal clarity.
- Easy-to-read graduations for accurate volume determination.
- Pipets are color-coded based on sizes for easy identification.
- Sterility assurance level of 10-6.
- Compatible with all mechanical and electronic pipette controllers - including Pipet Helper® or Easypet® 3.
- Free of detectable pyrogens, DNA, RNase and DNase and certified non-cytotoxic.
Application
- Transfer of liquids
- Mixing suspensions
- Combining reagents and chemical solutions
- Transferring cells for empirical analysis or expansion
- Layering reagents for creating higher density gradients
Legal Information
Easypet is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Pipet Helper is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)