Skip to Content
Merck
All Photos(1)

Documents

EMU088231

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Yap1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CACACTGGAAGGAGATGCAATGAACATAGAAGGGGAGGAGCTGATGCCCAGTCTGCAGGAAGCGCTGAGTTCCGAAATCTTGGACGTGGAGTCTGTGTTGGCTGCCACCAAGCTAGATAAAGAAAGCTTTCTCACGTGGTTATAGAGCTGCAGGGAGCCACTCTGAGTCTGTGAGGGATCCACAGAGCCTAAGATGTGCACGCCTGAAATTCAGATAAGTCAGTGGGGGTTCTCTGGCTAACACAGAAAACAGATGAACCAGTGTCCATCGTTGTTCCGCTTTTCTCTGCCCGTCGCTGCTCTTACGTTGGTTGCTGACCTCTTCACGGCCGGCTCTAAAGAACCCGAACCGCAGACAGATTCCTTTGTTAACTCTGCTATGATAACTACGTTCTCTGGGATTGCTGGGGGATGGCCTGCTGGATAATGGATGTTCTGCCTTTTGTCCGGTGGTCCTTTCACCATCACTTTAACTGAACACACAGACTGGGAACTGAATGCTCTAGAACATTGTTCAAGAGGTGGTTTCTTCAGCTGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lise Lotte Christensen et al.
PloS one, 9(6), e96767-e96767 (2014-06-04)
MicroRNAs (miRNAs) play a critical role in many biological processes and are aberrantly expressed in human cancers. Particular miRNAs function either as tumor suppressors or oncogenes and appear to have diagnostic and prognostic significance. Although numerous miRNAs are dys-regulated in
Erica Lorenzetto et al.
Oncotarget, 5(9), 2608-2621 (2014-05-09)
The transcriptional coactivator YAP1 is a critical effector of the human Salvador-Warts-Hippo pathway. Literature data report apparently discrepant results on the carcinogenic role of YAP1, which acts either as oncogene or as tumor suppressor in different in vitro and in
David Fu et al.
Endocrine-related cancer, 21(2), 297-310 (2014-01-07)
The Hippo signaling pathway has been implicated as a conserved regulator of organ size in both Drosophila and mammals. Yes-associated protein (YAP), the central component of the Hippo signaling cascade, functions as an oncogene in several malignancies. Ovarian granulosa cell
C He et al.
Oncogene, 34(50), 6040-6054 (2015-03-24)
Mechanisms underlying ovarian cancer initiation and progression are unclear. Herein, we report that the Yes-associated protein (YAP), a major effector of the Hippo tumor suppressor pathway, interacts with ERBB signaling pathways to regulate the initiation and progression of ovarian cancer.
Hyun-Jung Choi et al.
Nature communications, 6, 6943-6943 (2015-05-13)
Angiogenesis is regulated by the dynamic interaction between endothelial cells (ECs). Hippo-Yes-associated protein (YAP) signalling has emerged as a key pathway that controls organ size and tissue growth by mediating cell contact inhibition. However, the role of YAP in EC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service