Skip to Content
Merck
All Photos(1)

Key Documents

EMU062931

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Vegfa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTACCTCCACCATGCCAAGTGGTCCCAGGCTGCACCCACGACAGAAGGAGAGCAGAAGTCCCATGAAGTGATCAAGTTCATGGATGTCTACCAGCGAAGCTACTGCCGTCCGATTGAGACCCTGGTGGACATCTTCCAGGAGTACCCCGACGAGATAGAGTACATCTTCAAGCCGTCCTGTGTGCCGCTGATGCGCTGTGCAGGCTGCTGTAACGATGAAGCCCTGGAGTGCGTGCCCACGTCAGAGAGCAACATCACCATGCAGATCATGCGGATCAAACCTCACCAAAGCCAGCACATAGGAGAGATGAGCTTCCTACAGCACAGCAGATGTGAATGCAGACCAAAGAAAGACAGAACAAAGCCAGAAAATCACTGTGAGCCTTGTTCAGAGCGGAGAAAGCATT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Ding et al.
Biomaterials, 35(25), 7214-7227 (2014-05-31)
We described here the mechanisms by which small interfering RNA (siRNA) molecules incorporated in reconstituted high density lipoprotein (rHDL) were efficiently transferred into the cytoplasm of cells to perform target-specific therapy of tumor angiogenesis. Using fluorescent-tagged apolipoprotein A-I (apoA-I) and
Xue-jun Shao et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 36(5), 2051-2062 (2015-07-24)
Down-expression of microRNA-497 (miR-497) was often found in malignancies. The purposes of this study were to determine the expression of miR-497 in human osteosarcoma and to establish the association between miR-497 expression with cell survival and the sensitivity to cisplatin
Zhili Wen et al.
PloS one, 9(9), e104666-e104666 (2014-09-25)
MicroRNAs have been appreciated in various cellular functions, including the regulation of angiogenesis. Mesenchymal-stem-cells (MSCs) transplanted to the MI heart improve cardiac function through paracrine-mediated angiogenesis. However, whether microRNAs regulate MSC induced angiogenesis remains to be clarified. Using microRNA microarray
Tingting Li et al.
International journal of nanomedicine, 10, 4279-4291 (2015-07-15)
Engineering a safe and high-efficiency delivery system for efficient RNA interference is critical for successful gene therapy. In this study, we designed a novel nanocarrier system of polyethyleneimine (PEI)-modified Fe3O4@SiO2, which allows high efficient loading of VEGF small hairpin (sh)RNA
Yinan Wang et al.
PloS one, 10(6), e0130341-e0130341 (2015-06-16)
The transcription factor Krüppel-like factor 4 (KLF4) has been implicated in regulating cell proliferation, migration and differentiation in a variety of human cells and is one of four factors required for the induction of pluripotent stem cell reprogramming. However, its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service