Skip to Content
Merck
All Photos(1)

Key Documents

EHU157781

Sigma-Aldrich

MISSION® esiRNA

targeting human FLNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACATCATCCGCAATGACAATGACACCTTCACGGTCAAGTACACGCCCCGGGGGGCTGGCAGCTACACCATTATGGTCCTCTTTGCTGACCAGGCCACGCCCACCAGCCCCATCCGAGTCAAGGTGGAGCCCTCTCATGACGCCAGTAAGGTGAAGGCCGAGGGCCCTGGCCTCAGTCGCACTGGTGTCGAGCTTGGCAAGCCCACCCACTTCACAGTAAATGCCAAAGCTGCTGGCAAAGGCAAGCTGGACGTCCAGTTCTCAGGACTCACCAAGGGGGATGCAGTGCGAGATGTGGACATCATCGACCACCATGACAACACCTACACAGTCAAGTACACGCCTGTCCAGCAGGGTCCAGTAGGCGTCAATGTCACTTATGGAGGGGATCCCATCCCTAAGAGCCCTTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Mingione et al.
Journal of molecular endocrinology, 58(2), 91-103 (2016-11-23)
Parathyroid tumors display reduced sensitivity to extracellular calcium ([Ca
Anshika Sharma et al.
Frontiers in microbiology, 11, 581867-581867 (2020-10-27)
Influenza A virus (IAV) poses a major threat to global public health and is known to employ various strategies to usurp the host machinery for survival. Due to its fast-evolving nature, IAVs tend to escape the effect of available drugs
Qian Wang et al.
PloS one, 10(4), e0123018-e0123018 (2015-04-11)
Polycystin-2 (PC2), encoded by the PKD2 gene, is mutated in ~15% of autosomal dominant polycystic kidney disease. Filamins are actin-binding proteins implicated in scaffolding and membrane stabilization. Here we studied the effects of filamin on PC2 stability using filamin-deficient human
E Peverelli et al.
Endocrinology, 155(8), 2932-2941 (2014-05-16)
Somatostatin receptor type 2 (SST2) is the main pharmacological target of medical therapy for GH-secreting pituitary tumors, but molecular mechanisms regulating its expression and signaling are largely unknown. The aim of this study was to investigate the role of cytoskeleton
Donatella Treppiedi et al.
Molecular and cellular endocrinology, 524, 111159-111159 (2021-01-12)
Somatostatin receptor type 5 (SST5) represents the main pharmacological target in the treatment of adrenocorticotroph hormone (ACTH)-secreting tumors. However, molecular predictors of responsiveness to pasireotide require further investigation. The cytoskeleton protein filamin A (FLNA) modulates the responsiveness to somatostatin analogs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service