Skip to Content
Merck
All Photos(1)

Key Documents

EHU151801

Sigma-Aldrich

MISSION® esiRNA

targeting human KLF4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGAAGGATCTCGGCCAATTTGGGGTTTTGGGTTTTGGCTTCGTTTCTTCTCTTCGTTGACTTTGGGGTTCAGGTGCCCCAGCTGCTTCGGGCTGCCGAGGACCTTCTGGGCCCCCACATTAATGAGGCAGCCACCTGGCGAGTCTGACATGGCTGTCAGCGACGCGCTGCTCCCATCTTTCTCCACGTTCGCGTCTGGCCCGGCGGGAAGGGAGAAGACACTGCGTCAAGCAGGTGCCCCGAATAACCGCTGGCGGGAGGAGCTCTCCCACATGAAGCGACTTCCCCCAGTGCTTCCCGGCCGCCCCTATGACCTGGCGGCGGCGACCGTGGCCACAGACCTGGAGAGCGGCGGAGCCGGTGCGGCTTGCGGCGGTAGCAACCTGGCGCCCCTACCTCGGAGAGAGACC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott Bell et al.
American journal of human genetics, 104(5), 815-834 (2019-04-30)
We identified individuals with variations in ACTL6B, a component of the chromatin remodeling machinery including the BAF complex. Ten individuals harbored bi-allelic mutations and presented with global developmental delay, epileptic encephalopathy, and spasticity, and ten individuals with de novo heterozygous
Bing Li et al.
Human gene therapy, 30(3), 339-351 (2018-09-13)
Kallistatin (KS) has been recognized as a plasma protein with anti-inflammatory functions. Macrophages are the primary inflammatory cells in atherosclerotic plaques. However, it is unknown whether KS plays a role in macrophage development and the pathogenesis of atherosclerosis. This study
Takuya Nagata et al.
Breast cancer (Tokyo, Japan), 24(2), 326-335 (2016-06-15)
Prognosis of breast cancer patients has been reported to depend on the expression of induced pluripotent stem (iPS) cell-inducing factors: KLF4 and NANOG. However, the relationship between KLF4 or NANOG expression in each breast cancer subtype and the life prognosis
Muhammad Mushtaq et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(27), 15673-15683 (2020-06-24)
Stemness encompasses the capability of a cell for self-renewal and differentiation. The stem cell maintains a balance between proliferation, quiescence, and regeneration via interactions with the microenvironment. Previously, we showed that ectopic expression of the mitochondrial ribosomal protein S18-2 (MRPS18-2)
Tran T Hien et al.
Journal of cellular physiology, 233(9), 7195-7205 (2018-03-27)
Diabetes is a major risk factor for cardiovascular disease and this is in part due to the effects of hyperglycemia on vascular smooth muscle cells. Small non-coding microRNAs are known to control smooth muscle phenotype and arterial contractility and are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service