Skip to Content
Merck
All Photos(1)

Key Documents

EHU145491

Sigma-Aldrich

MISSION® esiRNA

targeting human MEF2D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCACTGCCTACAACACAGATTACCAGTTGACCAGTGCAGAGCTCTCCTCCTTACCAGCCTTTAGTTCACCTGGGGGGCTGTCGCTAGGCAATGTCACTGCCTGGCAACAGCCACAGCAGCCCCAGCAGCCGCAGCAGCCACAGCCTCCACAGCAGCAGCCACCGCAGCCACAGCAGCCACAGCCACAGCAGCCTCAGCAGCCGCAACAGCCACCTCAGCAACAGTCCCACCTGGTCCCTGTATCTCTCAGCAACCTCATCCCGGGCAGCCCCCTGCCCCACGTGGGTGCTGCCCTCACAGTCACCACCCACCCCCACATCAGCATCAAGTCAGAACCGGTGTCCCCAAGCCGTGAGCGCAGCCCTGCGCCTCCCCCTCCAGCTGTGTTCCCAGCTGCCCGCCCTGAGCCTGGCGATGGTCTCAGCAGCCCAGCCGGGGGATCCTATGAGACGGGAGACCGGGATGACGGACGGGGGGACTTCGGGCCCACACTGGGCCTGCTGCGCCCAGCCCCAGAGCCTGAGGCTGAGGGCTCAGCTGTGAAGAGGATGCGGCTTGATA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuna Li et al.
Aging, 12(7), 6456-6466 (2020-04-10)
Cochlear ribbon synapses play a pivotal role in the prompt and precise acoustic signal transmission from inner hair cells (IHCs) to the spiral ganglion neurons, while noise and aging can damage ribbon synapses, resulting in sensorineural hearing loss. Recently, we
Yaru Dong et al.
Neurochemical research, 46(2), 299-308 (2020-11-13)
Parkinson's disease (PD) is a severe neurodegenerative disease characterized by selective loss of dopaminergic neurons, which reduces quality of life of patients and poses a heavy burden to the society. The pathological mechanism of PD remains unclear, and increasing efforts
Zhi-Qin Hu et al.
Oncotarget, 8(54), 92079-92089 (2017-12-02)
The role of microRNA-92b-3p (miR-92b-3p) in cardiac hypertrophy was not well illustrated. The present study aimed to investigate the expression and potential target of miR-92b-3p in angiotensin II (Ang-II)-induced mouse cardiac hypertrophy. MiR-92b-3p was markedly decreased in the myocardium of
Haiyun Chen et al.
Aging, 12(14), 14897-14917 (2020-07-28)
T-006, a new derivative of tetramethylpyrazine, has been recently found to protect against 6-hydroxydopamine (6-OHDA)-induced neuronal damage and clear α-synuclein (α-syn) by enhancing proteasome activity in an α-syn transgenic Parkinson's disease (PD) model. The effect of T-006 on the 1-methyl-4-phenyl-1
Jung-Hwa Han et al.
Life sciences, 135, 1-8 (2015-06-03)
bFGF is a potent mitogen of cells associated with fibrosis. Although ERK5 has been reported to play roles in the development of fibrosis, its roles in regulating bFGF-induced fibrotic responses are not understood, especially in lung fibroblasts. The authors investigated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service