Skip to Content
Merck
All Photos(1)

Key Documents

EHU093591

Sigma-Aldrich

MISSION® esiRNA

targeting human SIRT3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAAACTGGGAAGCTTGATGGACCAGACAAATAGGATGATGGCTGCCCCCACACAATAAATGGTAACATAGGAGACATCCACATCCCAATTCTGACAAGACCTCATGCCTGAAGACAGCTTGGGCAGGTGAAACCAGAATATGTGAACTGAGTGGACACCCGAGGCTGCCACTGGAATGTCTTCTCAGGCCATGAGCTGCAGTGACTGGTAGGGCTGTGTTTACAGTCAGGGCCACCCCGTCACATATACAAAGGAGCTGCCTGCCTGTTTGCTGTGTTGAACTCTTCACTCTGCTGAAGCTCCTAATGGAAAAAGCTTTCTTCTGACTGTGACCCTCTTGAACTGAATCAGACCAACTGGAATCCCAGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Application

MISSION® esiRNA has been used for transfection of cells to target SIRT3.

Biochem/physiol Actions

SIRT3 (sirtuin 3) is a mitochondrial deacetylase, which acts as an oncogene. It is associated with the initiation and progression of certain cancers. However, in some cases it also works anti-oncogenically. It plays a major role in mitochondrial activity via deacetylating proteins associated with energy metabolism, ATP generation, redox optimization, and mitochondrial biogenesis. It is also involved in transcription, insulin secretion and apoptosis. Deacetylation of cyclophilin-D via SIRT3 inhibits age-associated cardiac hypertrophy. SIRT3 also exhibits neuroprotective roles.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao Song et al.
Free radical biology & medicine, 112, 616-630 (2017-09-16)
Mitochondrial reactive oxygen species (ROS) production has been implicated in the pathogenesis of fluoride toxicity in liver. Melatonin, an indolamine synthesized in the pineal gland, was previously shown to protect against sodium fluoride (NaF)-induced hepatotoxicity. This study investigated the protective
Xiaokan Zhang et al.
Circulation, 137(19), 2052-2067 (2018-01-14)
Heart failure leads to mitochondrial dysfunction and metabolic abnormalities of the failing myocardium coupled with an energy-depleted state and cardiac remodeling. The mitochondrial deacetylase sirtuin 3 (SIRT3) plays a pivotal role in the maintenance of mitochondrial function through regulating the
SIRT3 is correlated with the malignancy of non-small cell lung cancer.
Xiong Y, et. al.
International Journal of Oncology, 50, 903-903 (2017)
Mitochondrial SIRT3 mediates adaptive responses of neurons to exercise and metabolic and excitatory challenges.
Cheng A
Cell Metabolism, 23, 128-142 (2016)
Marija Pinterić et al.
Antioxidants (Basel, Switzerland), 9(4) (2020-04-05)
Estrogen (E2) is a major risk factor for the initiation and progression of malignancy in estrogen receptor (ER) positive breast cancers, whereas sirtuin 3 (Sirt3), a major mitochondrial NAD+-dependent deacetylase, has the inhibitory effect on the tumorigenic properties of ER

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service