Skip to Content
Merck
All Photos(1)

Key Documents

EHU089561

Sigma-Aldrich

MISSION® esiRNA

targeting human ADAM12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACCTCTGGATCCCAGTGAAGAGCTTCGACTCCAAGAATCATCCAGAAGTGCTGAATATTCGACTACAACGGGAAAGCAAAGAACTGATCATAAATCTGGAAAGAAATGAAGGTCTCATTGCCAGCAGTTTCACGGAAACCCACTATCTGCAAGACGGTACTGATGTCTCCCTCGCTCGAAATTACACGGTAATTCTGGGTCACTGTTACTACCATGGACATGTACGGGGATATTCTGATTCAGCAGTCAGTCTCAGCACGTGTTCTGGTCTCAGGGGACTTATTGTGTTTGAAAATGAAAGCTATGTCTTAGAACCAATGAAAAGTGCAACCAACAGATACAAACTCTTCCCAGCGAAGAAGCTGAAAAGCGTCCGGGGATCATGTGGATCACATCACAACACACCAAACCTCGCTGCAAAGAATGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingdan Chen et al.
Experimental biology and medicine (Maywood, N.J.), 242(14), 1432-1443 (2017-06-24)
Individuals with diabetes mellitus suffer from impaired angiogenesis and this contributes to poorer peripheral arterial disease outcomes. In experimental peripheral arterial disease, angiogenesis and perfusion recovery are impaired in mice with diabetes. We recently showed that a disintegrin and metalloproteinase
Junwen Wang et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 97, 1066-1077 (2017-11-16)
Pituitary adenomas are the second most common primary brain tumor with invasive properties. We have previously identified that ADAM12 (a disintegrin and metalloprotease 12) overexpression is associated with the tumor invasion of pituitary adenomas, however, the underlying mechanism remains unknown.
Sara Duhachek-Muggy et al.
Molecular cancer, 16(1), 32-32 (2017-02-06)
ADAM12 is upregulated in human breast cancers and is a predictor of chemoresistance in estrogen receptor-negative tumors. ADAM12 is induced during epithelial-to-mesenchymal transition, a feature associated with claudin-low breast tumors, which are enriched in cancer stem cell (CSC) markers. It
Yuto Nakamura et al.
American journal of physiology. Heart and circulatory physiology, 318(2), H238-H251 (2019-11-28)
A disintegrin and metalloproteinase (ADAM)12 is considered to promote cardiac dysfunction based on the finding that a small-molecule ADAM12 inhibitor, KB-R7785, ameliorated cardiac function in a transverse aortic constriction (TAC) model by inhibiting the proteolytic activation of heparin-binding-EGF signaling. However
Roopali Roy et al.
Molecular cancer research : MCR, 15(11), 1608-1622 (2017-08-03)
ADAM12, (

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service