Skip to Content
Merck
All Photos(1)

Key Documents

EHU018231

Sigma-Aldrich

MISSION® esiRNA

targeting human SP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGCTCCCAACTTACAGAACCAGCAAGTTCTGACAGGACTACCTGGAGTGATGCCTAATATTCAGTATCAAGTAATCCCACAGTTCCAGACCGTTGATGGGCAACAGCTGCAGTTTGCTGCCACTGGGGCCCAAGTGCAGCAGGATGGTTCTGGTCAAATACAGATCATACCAGGTGCAAACCAACAGATTATCACAAATCGAGGAAGTGGAGGCAACATCATTGCTGCTATGCCAAACCTACTCCAGCAGGCTGTCCCCCTCCAAGGCCTGGCTAATAATGTACTCTCAGGACAGACTCAGTATGTGACCAATGTACCAGTGGCCCTGAATGGGAACATCACCTTGCTACCTGTCAACAGCGTTTCTGCAGCTACCTTGACTCCCAGCTCTCAGGCAGTCACGATCAGCAGCTCTGGGTCCCAGGAGAGTGGCTCACAGCCTGTCACCTCAGGGACTACCATCAGTTCTGCCAGCTTGGTATCATCACAAGCCAGTTCCAGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Qi et al.
Journal of diabetes research, 2020, 2308520-2308520 (2020-03-19)
Cyclooxygenase 2 (COX2) and inducible nitric oxide synthase (iNOS) overexpression results in endothelial apoptosis, thus mediating vascular endothelial injury in hyperglycaemia. E26 transformation-specific sequence transcription factor-1 (ESE-1), which belongs to the E26 transformation-specific family of transcription factors, has been demonstrated
Lili Lv et al.
Oncology research, 26(5), 775-783 (2017-12-16)
Cervical cancer is the third most commonly diagnosed malignancy and the fourth leading cause of cancer-related deaths in women worldwide. MicroRNA-296 (miR-296) is aberrantly expressed in a variety of human cancer types. However, the expression levels, biological roles, and underlying
Xiaowei Diao et al.
Oncology reports, 40(2), 1174-1184 (2018-06-20)
Erythropoietin‑producing hepatocellular carcinoma cell surface type‑A receptor 3 (EPHA3) has been found to promote the proliferation and survival of prostate cancer (PCa) cell lines and prostate tumor development in nude mice. However, the regulation of EPHA3 in PCa remains largely unknown.
Im-Kyung Kim et al.
Scientific reports, 9(1), 5933-5933 (2019-04-13)
Specific protein 1 (SP1) is associated with aggressive behavior, invasive clinical phenotype and poor clinical outcomes in various cancers. We studied whether SP1 exerts its effect on invasiveness and promotion of the epithelial-mesenchymal transition (EMT) by regulating lysyl oxidase-like 2
Qiang Cai et al.
American journal of translational research, 8(10), 4068-4081 (2016-11-11)
Gallbladder cancer (GBC) is one of the most lethal cancers with poor prognosis. In this study, we report that the long non-coding RNA LINC00152 is significantly upregulated in GBC tissues and cell lines. The high LINC00152 levels correlated positively with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service