Skip to Content
Merck
All Photos(3)

Key Documents

E8126

Sigma-Aldrich

Equilin

Synonym(s):

1,3,5(10),7-Estratetraen-3-ol-17-one, 3-Hydroxy-1,3,5(10),7-estratetraen-17-one, 7-Dehydroestrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20O2
CAS Number:
Molecular Weight:
268.35
Beilstein:
2624302
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

sterility

non-sterile

Assay

≥98% (HPLC)

form

powder

technique(s)

inhibition assay: suitable

mp

238-240 °C (lit.)

solubility

chloroform: methanol (1:1): 40 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

C[C@]12CC[C@H]3C(=CCc4cc(O)ccc34)[C@@H]1CCC2=O

InChI

1S/C18H20O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3-5,10,14,16,19H,2,6-9H2,1H3/t14-,16+,18+/m1/s1

InChI key

WKRLQDKEXYKHJB-HFTRVMKXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Equilin is the Mare estrogen and is majorly used in multi-estrogen drugs for hormone replacement therapy. It is synthesized in the microsomes of mare placenta. It mediates estrogen receptor β (ERβ) expression resulting in the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB) which favors monocytes adhesion in atherosclerosis. In vitro equilin inhibits 17β-hydroxysteroid dehydrogenase type 1 (17β-HSD1) that catalyzes the conversion of estrone to 17β-estradiol.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Equilin and equilenin biosynthesis. Stereochemistry of aromatization of 3-hydroxy-3, 5, 7-androstatrien-17-one by horse placenta
Mitsuteru N and Yoshio O
Journal of Steroid Biochemistry, 26(1), 137-143 (1987)
Structure of the ternary complex of human 17beta-hydroxysteroid dehydrogenase type 1 with 3-hydroxyestra-1, 3, 5, 7-tetraen-17-one (equilin) and NADP+
Sawicki MW, et al.
Proceedings of the National Academy of Sciences of the USA, 96(3), 840-845 (1999)
Equilin
Handbook of Hormones, 582-582 (2016)
Naomi Suzuki et al.
Biochemistry, 43(35), 11312-11320 (2004-09-16)
Estrogen replacement therapy (ERT), composed of equilenin, is associated with increased risk of breast, ovarian, and endometrial cancers. Several diastereoisomers of unique dC and dA DNA adducts were derived from 4-hydroxyequilenin (4-OHEN), a metabolite of equilenin, and have been detected
Ekaterina V Rokhina et al.
The Science of the total environment, 417-418, 280-290 (2012-01-17)
Estrone (E1), 17β-estradiol (E2), estriol (E3), equilin (EQ) and 17α-estradiol (17α) estrogen hormones are released by humans and animals and have been detected in the environment and municipal wastewater treatment plants. The structural and electronic properties of natural hormone molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service