Skip to Content
Merck
All Photos(1)

Key Documents

E8031

Sigma-Aldrich

Esmolol hydrochloride

≥98% (HPLC), solid

Synonym(s):

4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzenepropanoic acid methyl ester hydrochloride hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H26NClO4
CAS Number:
Molecular Weight:
331.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

mp

89.0-90.6 °C

solubility

H2O: 12 mg/mL

originator

Baxter

storage temp.

2-8°C

SMILES string

Cl.COC(=O)CCc1ccc(OCC(O)CNC(C)C)cc1

InChI

1S/C16H25NO4.ClH/c1-12(2)17-10-14(18)11-21-15-7-4-13(5-8-15)6-9-16(19)20-3;/h4-5,7-8,12,14,17-18H,6,9-11H2,1-3H3;1H

InChI key

GEKNCWBANDDJJL-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Esmolol hydrochloride (ESM) may be used as a standard in spectrophotometric and electrochemical method for the determination of ESM in biological samples and pharmaceutical formulations.

Biochem/physiol Actions

Esmolol hydrochloride (ESM) is a class II antiarrhythmic drug. It is used to decrease the force and rate of heart contractions during surgery. Overdose of ESM can lead to side effects such as bradycardia, cardiac arrest, hypoventilation, mood disorder, and coma.
Esmolol hydrochloride is a ultrashort-acting β1 adrenoceptor antagonist (β1 receptor occupancy = 84.7%).

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Baxter. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of esmolol hydrochloride using analytical reagents
Narayana A, et al.
Indian Journal of Advances in Chemical Science null
Electrochemical System for Determination of Esmolol Hydrochloride Using Square Wave Adsorptive Stripping Voltammetry
Alarfaj NA and El-Tohamy MF
International Journal of Electrochemical Science (2016)
George Dimopoulos et al.
The Journal of surgical research, 198(1), 175-184 (2015-06-16)
Based on previous animal studies showing promising immunomodulatory efficacy esmolol, a selective β1-blocker, it was assumed that administration of esmolol in experimental pyelonephritis by multidrug-resistant Pseudomonas aeruginosa would prolong survival and modulate immune response. Acute pyelonephritis was induced in 80
Yong-Feng Zhu et al.
Journal of laparoendoscopic & advanced surgical techniques. Part A, 25(11), 903-907 (2015-11-18)
To determine the effect of lavage with adrenaline solution on CO2 absorption during retroperitoneal laparoscopic surgery. Sixty patients scheduled to undergo retroperitoneal laparoscopic surgery were divided into an AD group (lavage with normal saline containing adrenaline [1:500,000], n = 30)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service