Skip to Content
Merck
All Photos(1)

Key Documents

E7384

Sigma-Aldrich

[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt

≥97% (HPLC)

Synonym(s):

DAGO, DAMGO, Tyr-D-Ala-Gly-N-methyl-Phe-Gly-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H35N5O6
CAS Number:
Molecular Weight:
513.59
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(O)=O.C[C@@H](NC(=O)[C@@H](N)Cc1ccc(O)cc1)C(=O)NCC(=O)N(C)[C@@H](Cc2ccccc2)C(=O)NCCO

InChI

1S/C26H35N5O6.C2H4O2/c1-17(30-25(36)21(27)14-19-8-10-20(33)11-9-19)24(35)29-16-23(34)31(2)22(26(37)28-12-13-32)15-18-6-4-3-5-7-18;1-2(3)4/h3-11,17,21-22,32-33H,12-16,27H2,1-2H3,(H,28,37)(H,29,35)(H,30,36);1H3,(H,3,4)/t17-,21+,22+;/m1./s1

InChI key

XZZYKCKUDLGXJA-NJUGUJQKSA-N

Gene Information

human ... OPRM1(4988)
mouse ... OPRM1(18390)
rat ... OPRM1(25601)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Amino Acid Sequence

Tyr-Ala-Gly-Nme-Phe-Gly-ol

General description

[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt or DAMGO is a full agonist of the μ-opioid receptor. μ-opioid receptor is encoded by the OPRM1 gene, which has a predominant expression in reward-processing areas of brain. It acts as a receptor for endogenous opioids such as β-endorphin, encephalin, as well as foreign opioids such as morphine, heroin, and methadone.

Application

[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt or DAMGO has been used:
  • for use as a positive control in the assay of G-protein activation with μ-opioid receptor, to assay the inhibition of cAMP inhibition by DAMGO
  • for the determination of the suppression of contraction by opioid receptors
  • to determine whether the peripheral application of DAMGO suppresses the hypertonic saline (HS)-induced masseter nociception in slightly anaesthetized rats

Biochem/physiol Actions

Enkephalin analog that is a selective agonist at μ-opioid receptors.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Driessen et al.
British journal of pharmacology, 108(2), 443-447 (1993-02-01)
1. Effects of opioid agonists on the purinergic and adrenergic components of neurogenic contractions and in some experiments on transmitter overflow were studied in the mouse isolated vas deferens. 2. When the vas deferens was stimulated every 2 min by
S Nũnéz et al.
Neuroscience, 146(3), 1346-1354 (2007-03-24)
The aims of this project were to investigate whether inflammation in the orofacial muscle alters mu opioid receptor (MOR) mRNA and protein expressions in trigeminal ganglia (TG), and to assess the contribution of peripheral MORs under acute and inflammatory muscle
Frank J Meye et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(46), 16120-16128 (2012-11-16)
μ-Opioid receptors (MORs) in the ventral tegmental area (VTA) are pivotally involved in addictive behavior. While MORs are typically activated by opioids, they can also become constitutively active in the absence of any agonist. In the current study, we present
Javier Llorente et al.
The European journal of neuroscience, 36(12), 3636-3642 (2012-09-26)
There is considerable controversy over whether μ-opioid receptor (MOPr) desensitization is homologous or heterologous and over the mechanisms underlying such desensitization. In different cell types MOPr desensitization has been reported to involve receptor phosphorylation by various kinases, including G-protein-coupled receptor
Ken W K Lee et al.
Journal of psychiatry & neuroscience : JPN, 40(1), 38-45 (2014-10-01)
Preference for fatty foods is a risk factor for obesity. It is a complex behaviour that involves the brain reward system and is regulated by genetic and environmental factors, such as the opioid receptor mu-1 gene (OPRM1) and prenatal exposure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service