Skip to Content
Merck
All Photos(3)

Key Documents

E3289

Sigma-Aldrich

Catalase from bovine liver

≥20000 units/mg protein, lyophilized powder

Synonym(s):

H2O2:H2O2 oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥20000 units/mg protein

mol wt

tetramer ~250 kDa

composition

protein, ≥60% E405

isoelectric point

5.4

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C9H10O3/c1-2-12-9(11)7-3-5-8(10)6-4-7/h3-6,10H,2H2,1H3

InChI key

NUVBSKCKDOMJSU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

Catalase is encoded by the gene mapped to human chromosome 11p13. Mammalian catalase is an antioxidant enzyme and is a member of hydroperoxidase family). The active enzyme is homotetrameric in nature.
Research Area: Cell Signaling

Catalase from bovine liver is a tetramer consisting of 4 equal subunits each with a 60 kDa molecular weight. Each of these subunits contains iron bound to a protoheme IX group. The enzyme will also strongly bind to NADP, where NADP and the heme group are within 13.7 angstroms.

Application

Catalase from bovine liver has been used:
  • as a component of gloxy, which is one of the most popular oxygen-scavenging systems.
  • in single molecule fluorescence resonance energy transfer (FRET) measurements.
  • in cytochrome-C oxidase (COX) histochemistry to study the role of mitochondrial uncoupling protein 2 (UCP2) in limiting oxidative stress during glaucoma.
  • to pre-treat slide chambers for total internal reflection fluorescence (TIRF) microscopy.
  • in quantifying hydrogen peroxide release.
  • to help minimize the effects of reactive oxygen species (ROS) in cooperativity assay for studying doublecortin and microtubule interaction.
Catalase acts as a natural antioxidant to study the roles of reactive oxygen species in gene expression and apoptosis. It has also been used to protect against oxidative damage to proteins, lipids, and nucleic acids. Industrially, catalzes have been used to remove hydrogen peroxide added to milk and cheese, in textile bleaching, and to examine its positive effects on the viability of DNA-repair mutants of E. coli.

Catalase from bovine liver may be used:

  • to prepare H2O2-O2 based biocathode for applications in glucose biofuel cells
  • to study the kinetic properties and storage stability of catalase immobilized on to florisil
  • in glutathione-mediated superoxide generation in an aqueous solution

Biochem/physiol Actions

Catalase has an ability to convert solar radiation into reactive oxidant species (ROS). It is involved in regulation of tissue peroxide levels. Mutation in catalase gene result in the progress of essential hypertension (EH).
Catalase, an antioxidant enzyme found in all aerobic organisms, catalyzes the degradation of hydrogen peroxide, a byproduct of metabolic processes, into less harmful water and oxygen. It can also react with alkylhydrogen peroxides, such as methylperoxide and ethylperoxide and the second H2O2 molecule can be replaced by methanol, ethanol, propanol, formate and nitrate as a hydrogen donor. Catalase enzyme uses either iron (Fe) or manganese (Mn) as cofactor, and are classified as Fe-CAT or Mn-CAT.

Unit Definition

One unit will decompose 1.0 micromole of hydrogen peroxide per minute at pH 7.0 at 25 deg C, while the hydrogen peroxide concentration falls from 10.3 to 9.2 millimolar.

Physical form

Supplied as a lyophilized powder containing ≤0.2% thymol

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms of oxidant generation by catalase.
Heck D E, et al.
Annals of the New York Academy of Sciences, 1203(1), 120-125 (2010)
Observation and Analysis of RAD51 Nucleation Dynamics at Single-Monomer Resolution.
Subramanyam S, et al.
Methods in Enzymology, 600, 201-232 (2018)
Frequent loss of heterozygosity on chromosomes 3p and 17p without VHL or p53 mutations suggests involvement of unidentified tumor suppressor genes in follicular thyroid carcinoma.
Grebe S K, et al.
The Journal of Clinical Endocrinology and Metabolism, 82(11), 3684-3691 (1997)
Influence of A-21T and C-262T genetic polymorphisms at the promoter region of the catalase (CAT) on gene expression.
Saify K, et al.
Environmental Health and Preventive Medicine, 21(5), 382-386 (2016)
The yeast INO80 complex operates as a tunable DNA length-sensitive switch to regulate nucleosome sliding
Zhou CY, et al.
Molecular Cell, 69(4), 677?688-677?688 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service