Skip to Content
Merck
All Photos(1)

Key Documents

E1388

Sigma-Aldrich

β-Endothelial Cell Growth Factor human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

ECGS, β-ECGF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

β-Endothelial Cell Growth Factor human, β-ECGF, recombinant, expressed in E. coli, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

0.03-0.3 ng/mL ED50/EC50

quality

endotoxin tested

mol wt

predicted mol wt ~17 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles (Do not store in a frost-free freezer.)

technique(s)

cell culture | mammalian: suitable

impurities

≤1.0 EU/μg protein Endotoxin level (LAL test)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... VEGFB(7423)

General description

The β-endothelial cell growth factor (b-ECGF) gene encodes an angiogenic factor, and is mapped to human chromosome 11q13.1. Recombinant, human b-endothelial cell growth factor (b-ECGF) is a 14 amino acid N-terminal extension of acidic fibroblast growth factor. It is expressed in Escherichia coli. The 154 amino acid recombinant protein, generated after removal of the initiation methionine, has a predicted molecular mass of about 17 kDa.

Application

β-Endothelial Cell Growth Factor human has been used:
  • in cell death assay
  • in endothelial cell culture
  • in human umbilical vascular endothelial cell (HUVEC) culture

Biochem/physiol Actions

β-Endothelial cell growth factor (β-ECGF) is a potent endothelial cell mitogen and its mitogenic activity is potentiated by heparin. β-ECGF stimulates the growth of quiescent HUVEC cells in culture. β-ECGF is associated with lipid and glucose metabolism. This growth factor induces neovascularization in disease conditions. Upregulation of β-ECGF is observed in diabetic retinopathy, and polycystic ovary syndrome. It is also associated with tumor proliferation and invasion. β-ECGF increases fatty acid transport proteins expression to improve endothelial mediated lipid transpor.

Other Notes

14 amino acid, N-terminal extended form of human FGF-acidic.

Physical form

Lyophilized from a 0.2 μm filtered solution in 10 mM MOPS, 50 mM ammonium sulfate, 0.2 mM DTT, and 0.2 mM EDTA, pH 7.2, containing 50 μg bovine serum albumin per 1 μg cytokine.

Analysis Note

The proliferative activity is tested in culture by using quiescent NR6R-3T3 fibroblasts.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eleanor G Lynam et al.
Methods in molecular biology (Clifton, N.J.), 2475, 223-228 (2022-04-23)
Endothelial cell proliferation rate is an important indicator of vascular health. Being able to detect the rate of endothelial cell proliferation, or cell cycle disturbances after intervention is a valuable tool for analysing any beneficial or detrimental effects of treatments
Serum vascular endothelial growth factor B is elevated in women with polycystic ovary syndrome and can be decreased with metformin treatment
Cheng F, et al.
Clinical Endocrinology, 84(3), 386-393 (2016)
A blood pressure-associated variant of the SLC39A8 gene influences cellular cadmium accumulation and toxicity
Zhang R, et al.
Human Molecular Genetics, 25(18), 4117-4126 (2016)
Chuan-Rong Zhao et al.
Small (Weinheim an der Bergstrasse, Germany), 19(2), e2204694-e2204694 (2022-11-21)
Disturbed blood flow induces endothelial pro-inflammatory responses that promote atherogenesis. Nanoparticle-based therapeutics aimed at treating endothelial inflammation in vasculature where disturbed flow occurs may provide a promising avenue to prevent atherosclerosis. By using a vertical-step flow apparatus and a microfluidic
Plasma vascular endothelial growth factor B levels are increased in patients with newly diagnosed type 2 diabetes mellitus and associated with the first phase of glucose-stimulated insulin secretion function of beta-cell
Wu J, et al.
Journal of Endocrinological Investigation, 40(11), 1219-1226 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service