Skip to Content
Merck
All Photos(2)

Key Documents

E1281

Sigma-Aldrich

Anti-Ezrin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-80K, Anti-Cytovillin, Anti-p81

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

human, rat

technique(s)

immunohistochemistry: 20-40 μg/mL
immunoprecipitation (IP): 0.5-1 μg using RIPA extract of human A431 cells
indirect immunofluorescence: 10-20 μg/mL
western blot (chemiluminescent): 0.5-1 μg/mL using whole extract of rat NRK cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EZR(7430)
mouse ... Ezr(22350)
rat ... Ezr(54319)

General description

Ezrin is a 80kDa membrane-associated protein that belongs to ERM (Ezrin/Radixin/Moesin) family. Ezrin is found predominantly in polarized epithelial cells such as intestinal epithelial cells, gastric parietal cells, epithelial cells of the kidney proximal tubule, terminal bronchiole of the lung, and in mesothelia.

Immunogen

synthetic peptide corresponding to amino acids 479-498 of human ezrin with N-terminal added cysteine, conjugated to KLH. The corresponding sequence in rat and mouse differs by three amino acids.

Application

Anti-Ezrin antibody produced in rabbit has been used in:
  • immunolocalization studies
  • immunostaining
  • immunofluorescence and immunohistochemistry
  • immunoblot

Biochem/physiol Actions

Cultured neuronal cells do express ezrin to some extent, and whole brain tissue expresses low levels of ezrin. Ezrin expression is associated with malignancy in astrocytoma, osteosarcoma, mammary and pancreatic adenocarcinomas, and in cutaneous and uveal melanomas.
Ezrin promotes cell adhesion. The NH2-terminal domain negatively regulates membrane spreading and elongation properties of ezrin COOH-terminal domain. The members of ERM family are usually involved in cross-linking of plasma membrane and actin filaments. ERM proteins are involved in cell adhesion, cell shape determination, migration, and the organization of cell surface structures. They are also involved in signal transduction and apoptosis. Ezrin has been implicated in tumor progression and metastatic spread.

Physical form

Solution in 0.01 M phosphate buffered saline containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AICAR induces mitochondrial apoptosis in human osteosarcoma cells through an AMPK-dependent pathway
Morishita M, et al.
International journal of oncology, 50(1), 23-30 (2017)
K D Geiger et al.
The American journal of pathology, 157(6), 1785-1793 (2000-12-07)
The actin-binding protein ezrin has been associated with motility and invasive behavior of malignant cells. To assess the presence of this protein in human glial cells of the brain and its potential role in benign and malignant glial tumors, we
Heat shock protein family B member 1 facilitates ezrin activation to control cell migration in esophageal squamous cell carcinoma
Xie YH, et al.
The International Journal of Biochemistry & Cell Biology, 112(19), 79-87 (2019)
Jorge A Lobato-Álvarez et al.
Frontiers in physiology, 7, 450-450 (2016-10-25)
Na+, K+-ATPase, or the Na+ pump, is a key component in the maintenance of the epithelial phenotype. In most epithelia, the pump is located in the basolateral domain. Studies from our laboratory have shown that the β1 subunit of Na+
Suvi Ilmonen et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 18(4), 503-510 (2004-10-12)
Ezrin is a member of the ezrin-radixin-moesin family of proteins that link the actin-containing cytoskeleton to the plasma membrane. Ezrin is also connected to signaling molecules involved in the regulation of cell survival, proliferation and migration. Here, we examined the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service