Skip to Content
Merck
All Photos(1)

Documents

E1036

Sigma-Aldrich

116-9e

≥98% (HPLC), powder

Synonym(s):

116-9e, 4-[1,1′-Biphenyl]-4-yl-3,4-dihydro-6-methyl-2-oxo-5-[(phenylmethoxy)carbonyl]-1(2H)-pyrimidinehexanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H32N2O5
CAS Number:
Molecular Weight:
512.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

CC1=C(C(NC(=O)N1CCCCCC(O)=O)c2ccc(cc2)-c3ccccc3)C(=O)OCc4ccccc4

InChI

1S/C31H32N2O5/c1-22-28(30(36)38-21-23-11-5-2-6-12-23)29(32-31(37)33(22)20-10-4-9-15-27(34)35)26-18-16-25(17-19-26)24-13-7-3-8-14-24/h2-3,5-8,11-14,16-19,29H,4,9-10,15,20-21H2,1H3,(H,32,37)(H,34,35)

InChI key

GHFQWLNXJMUCGC-UHFFFAOYSA-N

Application

116-9e has been used as an inhibitor of heat shock protein 70 (Hsp70) co-chaperone HDJ2 (human Ydj1/DNAJA1) to study the influence of HDJ2 on the regulation of ribonucleotide reductase (RNR) activity in HEK293 cells. It has also been used as an Hsp70 inhibitor to study the effect of Hsp70 chaperone on b5‐ops glycosylation in the side populations (SP) HeLa cells.

Biochem/physiol Actions

116-9e is a blocker of Hsp40-Hsp70 binding thereby inhibiting the chaperone activity of Hsp70-Hsp40. The Hsp40 family of co–chaperones binds to Hsp70 through a conserved J–domain. It is believed that 116-9e inhibits chaperone functions by preventing Hsp70–Hsp40 complex assembly.

The same compound by a different name, MAL2-11B, has been found to inhibit the activity of a viral J-domain protein, large tumor antigen (TAg). MAL2-11B inhibited both TAg′s endogenous ATPase activity and the TAg-mediated activation of Hsp70.
116-9e is a dihhydropyrimidine compound.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bruna Figueiredo Costa et al.
Traffic (Copenhagen, Denmark), 19(3), 182-197 (2018-01-24)
Tail-anchored (TA) proteins insert into their target organelles by incompletely elucidated posttranslational pathways. Some TA proteins spontaneously insert into protein-free liposomes, yet target a specific organelle in vivo. Two spontaneously inserting cytochrome b5 forms, b5-ER and b5-RR, which differ only
Isaac T Sluder et al.
PLoS genetics, 14(11), e1007462-e1007462 (2018-11-20)
Hsp70 is a well-conserved molecular chaperone involved in the folding, stabilization, and eventual degradation of many "client" proteins. Hsp70 is regulated by a suite of co-chaperone molecules that assist in Hsp70-client interaction and stimulate the intrinsic ATPase activity of Hsp70.
Erina Matsuoka et al.
Functional plant biology : FPB (2019-06-21)
The heat shock protein 90 (HSP90) inhibitor, geldanamycin, is a chemical inducer of the heat shock response (HSR) in Arabidopsis. Geldanamycin is thought to activate the heat shock signal by dissociating the HSP90-heat shock factor (HSF) complex. Recent studies have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service