Skip to Content
Merck
All Photos(1)

Key Documents

E0502

Sigma-Aldrich

Elastin–Congo red

elastase substrate

Synonym(s):

Elastin Congo red

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

Quality Level

form

powder

storage temp.

2-8°C

General description

Elastin−Congo red is a substrate-dye conjugate and is used in agar medium to probe elastase activity. The presence of elastase functionality leads to cleavage of the dye from the elastin substrate.

Application

Elastin−Congo red has been used as a substrate for neutrophil elastase, salivary elastase and P. aeruginosa.

Other Notes

Elastin impregnated with Congo red dye.

Substrates

Substrate for the estimation of elastase.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of an agar diffusion method to measure elastase inhibition activity using Elastin-Congo red
Jung KH and Kim HJ
Journal of microbiology and biotechnology, 16(8), 1320-1324 (2006)
Salivary immunity in elderly individuals presented with Candida-related denture stomatitis
Gasparoto T, et al.
Gerodontology, 29(2), e331-e339 (2012)
Faizan Abul Qais et al.
ACS omega, 6(29), 18823-18835 (2021-08-03)
Quorum sensing (QS) and biofilm inhibition are recognized as the novel drug targets for the broad-spectrum anti-infective strategy to combat the infections caused by drug-resistant bacterial pathogens. Many compounds from medicinal plants have been found to demonstrate anti-infective activity. However
AiiM lactonase strongly reduces quorum sensing controlled virulence factors in clinical strains of Pseudomonas aeruginosa isolated from burned patients
Lopez-Jacome LE, et al.
Frontiers in Microbiology, 10, 2657-2657 (2019)
Activation pattern of neutrophils from blood of elderly individuals with Candida-related denture stomatitis
Gasparoto TH, et al.
European Journal of Clinical Microbiology & Infectious Diseases : Official Publication of the European Society of Clinical Microbiology, 31(6), 1271-1277 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service