Skip to Content
Merck
All Photos(1)

Key Documents

DUO82040

Sigma-Aldrich

Duolink® In Situ Mounting Medium with DAPI

Synonym(s):

in situ Proximity Ligation Assay reagent, Protein Protein Interaction Assay reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

product line

Duolink®

Quality Level

technique(s)

proximity ligation assay: suitable

fluorescence

λex 360 nm; λem 460 nm (Zeiss Filter set 49)

suitability

suitable for fluorescence

shipped in

wet ice

storage temp.

2-8°C

Application

Duolink® proximity ligation assay(PLA®) allows for endogenous detection of protein interactions, post translational modifications, and protein expression levels at the single molecule level in fixed cells and tissue samples.

Use the Duolink® In Situ Fluorescence Protocol for this product. A set of short instructionsis also available.

Visit our Duolink® PLA Resource Center for information on how to run a Duolink® experiment, applications, troubleshooting, and more.

To perform a complete Duolink®PLA in situ experiment you will need two primary antibodies (PLA, IHC, ICC or IF validated) that recognize two target epitopes. Other necessary reagents include a pair of PLA probes from different species (one PLUS and one MINUS), detection reagents, wash buffers, and mounting medium. Note that the primary antibodies must come from the same species as the Duolink® PLA probes. Analysis is carried out using standard immunofluorescence assay equipment.
Specificity
Duolink® In Situ Mounting Medium with DAPI is ideal for nuclear staining and preserving signals generated with the Duolink® In Situ Detection Reagents for fluorescence microscopy. See datasheet for more details.
Note: Counterstaining with Cy®2 is not recommended.

Application Note
Two primary antibodies raised in different species are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC) or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Duolink® in situ reagents are suitable for use on fixed cells, cytospin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). No minimum number of cells is required.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list
Duolink® In Situ mounting medium with DAPI has been used to mount coverslips in proximity ligation assay (PLA) for nuclear staining.[1][2][3]

Features and Benefits

  • No overexpression or genetic manipulation required
  • High specificity (fewer false positives)
  • Single molecule sensitivity due to rolling circle amplification
  • Relative quantification possible
  • No special equipment needed
  • Quicker and simpler than FRET
  • Increased accuracy compared to co-IP
  • Publication-ready results

Legal Information

Cy is a registered trademark of Cytiva
Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intraneuronal Amylin Deposition, Peroxidative Membrane Injury and Increased IL-1β Synthesis in Brains of Alzheimer?s Disease Patients with Type-2 Diabetes and in Diabetic HIP Rats
Verma N, et al.
Journal of Alzheimer'S Disease, 53(1), 259-272 (2016)
Detection of Receptor Heteromerization Using In Situ Proximity Ligation Assay
Gomes I, et al.
Current Protocols in Pharmacology / Editorial Board, S.J. Enna (editor-in-chief) ... [Et al.], 2-16 (2016)
S6K2-mediated regulation of TRBP as a determinant of miRNA expression in human primary lymphatic endothelial cells
Warner MJ, et al.
Nucleic Acids Research, 44(20), 9942-9955 (2016)
Takashi Miyamoto et al.
The Journal of biological chemistry, 291(4), 1719-1734 (2015-11-22)
Diverse lines of evidence suggest that amyloid-β (Aβ) peptides causally contribute to the pathogenesis of Alzheimer disease (AD), the most frequent neurodegenerative disorder. However, the mechanisms by which Aβ impairs neuronal functions remain to be fully elucidated. Previous studies showed
M Aubele et al.
British journal of cancer, 103(5), 663-667 (2010-08-12)
Protein tyrosine kinase 6 (PTK6; breast tumour kinase) is overexpressed in up to 86% of the invasive breast cancers, and its association with the oncoprotein human epidermal growth factor receptor 2 (HER2) was shown in vitro by co-precipitation. Furthermore, expression

Questions

  1. What is the refractive index of the Duolink® In Situ Mounting Medium with DAPI (DUO82040)? Is it compatible with Total Internal Reflection Fluorescence (TIRF) Microscopy?

    1 answer
    1. The refractive index of the Duolink® In Situ Mounting Medium with DAPI is 1.461. Compatibility with Total Internal Reflection Fluorescence (TIRF) Microscopy has not been tested in-house.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service