Skip to Content
Merck
All Photos(1)

Documents

D9696

Sigma-Aldrich

Dronedarone hydrochloride

≥98% (HPLC)

Synonym(s):

N-{2-Butyl-3-[4-(3-dibutylaminopropoxy)benzoyl]benzofuran-5-yl}methanesulfonamide hydrochloride, Multaq, SR-33589B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H44N2O5S · HCl
CAS Number:
Molecular Weight:
593.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: 15 mg/mL, clear

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

Cl.CCCCN(CCCC)CCCOc1ccc(cc1)C(=O)c2c(CCCC)oc3ccc(NS(C)(=O)=O)cc23

InChI

1S/C31H44N2O5S.ClH/c1-5-8-12-29-30(27-23-25(32-39(4,35)36)15-18-28(27)38-29)31(34)24-13-16-26(17-14-24)37-22-11-21-33(19-9-6-2)20-10-7-3;/h13-18,23,32H,5-12,19-22H2,1-4H3;1H

InChI key

DWKVCQXJYURSIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dronedarone hydrochloride has been used as an anti-arrhythmic drug to assess its impact in P19C5 embryoid bodies (EBs). It has also been used to determine its antiviral potency.

Biochem/physiol Actions

Dronedarone is a Class III antiarrhythmic and a multi-channel blocker for atrial fibrillation. It blocks potassium, sodium, and calcium channels and also exhibits antiadrenergic properties.
Dronedarone is a synthetic derivative of amiodarone, which belongs to the class of benzofurane anti-arrhythmic compounds. It might moderately block cytochrome P450 3A4 (CYP3A4) activity. Dronedarone is used to treat atrial flutter.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeremy A Schafer et al.
Cardiovascular therapeutics, 28(1), 38-47 (2010-01-16)
Atrial fibrillation (AF) is the most common sustained arrhythmia, affecting more than 2.2 million Americans. ACC/AHA/ESC guidelines for the management of patients with AF recommend amiodarone for maintaining sinus rhythm. Dronedarone is a derivative of amiodarone indicated for the treatment
Use of in vitro morphogenesis of mouse embryoid bodies to assess developmental toxicity of therapeutic drugs contraindicated in pregnancy
Warkus ELL, et al.
Toxicological Sciences, 149(1), 15-30 (2016)
Dronedarone: current evidence and future questions
Schafer JA, et al.
Cardiovascular Therapeutics, 28(1), 38-47 (2010)
Filovirus antiviral activity of cationic amphiphilic drugs is associated with lipophilicity and ability to induce phospholipidosis
Gunesch AP, et al.
Antimicrobial Agents and Chemotherapy (2020)
Stress degradation studies of dronedarone in pharmaceutical dosage form by a validated stability-indicating LC method
Ahirrao VK, et al.
Journal of the Chilean Chemical Society, 57(3), 1272-1276 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service