Skip to Content
Merck

D8281

Anti-Desmin antibody produced in rabbit

whole antiserum

enhanced validation

Enhanced Validation

Synonym(s):

Anti-Desmin, Desmin Detection Antibody, Rabbit Anti-Desmin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, WB
Citations:
63
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

chicken, bovine, human, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

indirect immunofluorescence: 1:20 using cultured cells, indirect immunofluorescence: 1:20 using human skeletal muscle sections, western blot: 1:100 using animal or human skeletal muscle.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DES(1674)
mouse ... Des(13346)

General description

Desmin is an intermediate filament protein found mainly in skeletal, cardiac, and smooth muscle cells. It acts as a marker for tumor origin. It also facilitates mechanical support for muscles, mitochondrial localization, myofibrillogenesis, gene expression regulation, and intracellular signalling. Anti-desmin antibody can be used for immunocytochemical localization of intermediate filaments in all types of muscle cells. It can also be used to stain a wide desmin band of 50,000 to 55,000 molecular weight using immunoblotting assays. Rabbit anti-desmin antibody reacts specifically with desmin in bovine, mouse, chicken and human.
Desmin is mapped to human chromosome 2q35. The desmin protein comprises of a head domain, four α-helical segments, three non-helical rod segments and a tail domain. It exists as dimer and tetramer assemblies. In smooth and striated muscles, desmin is localized in the periphery of Z-disk.

Immunogen

desmin from chicken gizzard

Application

Anti-Desmin antibody produced in rabbit has been used in:
  • immunohistochemistry of zebrafish intestines sections
  • immunodetection in fish embryo samples
  • immunolabeling of myofibers

Anti-desmin antibody can be used in immunocytochemistry and western immunoblotting. It can also be used in indirect immunofluorescence.

Biochem/physiol Actions

Desmin plays a crucial role in providing mechanical strength to muscle tissues. Mutations in desmin gene contributes to imbalance in skeletal proteins, resulting in desminopathies. Missense mutations in desmin gene leads to hypertrophic cardiomyopathy and systemic myopathy. Mutations in desmin gene impairs the stability of intermediary filaments in cardiomyocytes, contributing to pathophysiology of cardiac diseases.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library